Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
| Size/SKU | Availability | Price |
|---|---|---|
Available to ship TODAYfromMILWAUKEE | $620.00 |
About This Item
Empirical Formula (Hill Notation):
C48H46N12Na2O20P2
Molecular Weight:
1218.87
NACRES:
NA.77
UNSPSC Code:
51111800
Assay:
≥95% (HPLC)
Storage condition:
protect from light
Quality Segment
assay
≥95% (HPLC)
storage condition
protect from light
color
orange to dark orange-red
storage temp.
−20°C
Biochem/physiol Actions
cGAMPFluo is a fluorescein labeled analog of cyclic dinucleotide cGAMP, which after encapsulation can be used to study and verify internalization/delivery of cGAMP. Similarly to cGAMP it is not cell-permeable. λexc 494 nm and λem 517 nm.
fluorescein labeled analog of cyclic dinucleotide cGAMP
1 of 1
This Item | |||
|---|---|---|---|
| description ≥95% (HPLC) | description ≥98% (HPLC) | description 90-95% | description ≥95% (HPLC) |
| assay ≥95% (HPLC) | assay ≥98% (HPLC) | assay 90-95% | assay ≥95% (HPLC) |
| Quality Level 100 | Quality Level 100 | Quality Level 200 | Quality Level 100 |
| storage temp. −20°C | storage temp. −20°C | storage temp. −20°C | storage temp. −20°C |
| storage condition protect from light | storage condition - | storage condition - | storage condition - |
| color orange to dark orange-red | color white to beige | color - | color white to beige |
Storage Class
11 - Combustible Solids
wgk
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.



