Skip to Content
MilliporeSigma

Skip To

SML3175

BC1583

≥98% (HPLC)

Synonym(s):

1,1?-[1,4-Phenylenebis(oxy)]bis[3-[bis(phenylmethyl)amino]-2-propanol, 1-Dibenzylamino-3-[4-(3-dibenzylamino-2-hydroxy-propoxy)-phenoxy]-propan-2-ol

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
5 mg
Check Cart for Availability
$98.90
25 mg
Check Cart for Availability
$317.00

About This Item

Empirical Formula (Hill Notation):
C40H44N2O4
CAS Number:
Molecular Weight:
616.79
NACRES:
NA.77
UNSPSC Code:
12352200
Assay:
≥98% (HPLC)
Form:
powder

$98.90


Check Cart for Availability

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear (warmed)

storage temp.

room temp

Biochem/physiol Actions

BC1583 is a potent and selective Fbxo48 inhibitor that increases levels of pAmpkα in BEAS-2B cells. BC1583 potently stimulates Ampk-dependent signaling through inhibition of Fbxo48-mediated degradation of activated pAmpkα.
potent and selective Fbxo48 inhibitor that increases levels of pAmpkα in BEAS-2B cells

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
SML0989SML2682SML0515
description

≥98% (HPLC)

description

≥98% (HPLC)

description

≥98% (HPLC)

description

≥95% (HPLC)

form

powder

form

powder

form

powder

form

powder

assay

≥98% (HPLC)

assay

≥98% (HPLC)

assay

≥98% (HPLC)

assay

≥95% (HPLC)

Quality Level

100

Quality Level

100

Quality Level

-

Quality Level

100

storage temp.

room temp

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

room temp

solubility

DMSO: 2 mg/mL, clear (warmed)

solubility

DMSO: 25 mg/mL, clear

solubility

DMSO: 2 mg/mL, clear

solubility

DMSO: 5 mg/mL, clear (warmed)

color

white to beige

color

white to beige

color

faint brown to very dark brown-red

color

white to beige


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters