Skip to Content
MilliporeSigma

Skip To

SML3178

CPS2

≥98% (HPLC)

Synonym(s):

4-((5-amino-1-(3-methylthiophene-2-carbonyl)-1H-1,2,4-triazol-3-yl)amino)-N-(2-((1-(2-(2-(2-((2-(2,6-dioxopiperidin-3-yl)-1,3-dioxoisoindolin-4-yl)amino)ethoxy)ethoxy)ethyl)-1H-1,2,3-triazol-4-yl)methoxy)ethyl)benzenesulfonamide

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
5 mg

Estimated to ship TODAYfromMILWAUKEE

$226.00
25 mg

Estimated to ship TODAYfromMILWAUKEE

$905.00

About This Item

Empirical Formula (Hill Notation):
C38H42N12O10S2
CAS Number:
Molecular Weight:
890.94
NACRES:
NA.77
UNSPSC Code:
12352107
Assay:
≥98% (HPLC)
Form:
powder

$226.00


Estimated to ship TODAYDetails


​
Request a Custom Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

General description

CPS2 is derived from JNJ-7706621 which is a CDK/aurora inhibitor. It exhibits anti-proliferative effects.

Biochem/physiol Actions

CPS2 is a cell penetrant proteolysis-targeting chimera (PROTAC) that targets cyclin-dependent kinase 2 (CDK2). CPS2 potently degrades CDK2 in different cell lines. It appears that it does not induce comparable degradation of other targets
cell penetrant and selective proteolysis-targeting chimera (PROTAC) that targets cyclin-dependent kinase 2 (CDK2)

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
189406CPS260HPA040004
description

≥98% (HPLC)

description

The Aurora Kinase/Cdk Inhibitor, also referenced under CAS 443797-96-4, controls the biological activity of Aurora Kinase/Cdk. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.

description

sufficient for 60 mL substrate

description

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

form

powder

form

solid

form

liquid

form

buffered aqueous glycerol solution

assay

≥98% (HPLC)

assay

≥97% (HPLC)

assay

-

assay

-

Quality Level

100

Quality Level

100

Quality Level

100

Quality Level

100

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

−20°C

solubility

DMSO: 2 mg/mL, clear

solubility

DMSO: 15 mg/mL

solubility

-

solubility

-

color

white to beige

color

white

color

-

color

-


Storage Class

11 - Combustible Solids

WGK

WGK 3

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library




Questions

Reviews

No rating value

Active Filters