Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
| Size/SKU | Availability | Price |
|---|---|---|
5 mg | Available to ship TODAYfromMILWAUKEE | $167.00 |
25 mg | Available to ship TODAYfromMILWAUKEE | $534.00 |
About This Item
Empirical Formula (Hill Notation):
C18H17F2N3O2
CAS Number:
Molecular Weight:
345.34
UNSPSC Code:
12352200
NACRES:
NA.77
MDL number:
Assay:
≥98% (HPLC)
Form:
powder
Quality Segment
assay
≥98% (HPLC)
form
powder
color
white to beige
solubility
DMSO: 2 mg/mL, clear
storage temp.
2-8°C
SMILES string
FC1=C(C(F)=CC(C)=C1)C(C2=C(C3=C(N=C2)NN=C3)OCCCC)=O
InChI
1S/C18H17F2N3O2/c1-3-4-5-25-17-11(8-21-18-12(17)9-22-23-18)16(24)15-13(19)6-10(2)7-14(15)20/h6-9H,3-5H2,1-2H3,(H,21,22,23)
InChI key
SCFMWQIQBVZOQR-UHFFFAOYSA-N
Biochem/physiol Actions
ATP site-targeting, potent and CDK1/CDK2-selective cyclin-dependent kinase inhibitor with anti-cancer efficacy in A2780 ovarian cancer culture.
BMS-265246 is an ATP site-targeting, potent and CDK1/2-selective cyclin-dependent kinase inhibitor (IC50 = 6 nM/CDK1, 9 nM/CDK2, 230 nM/CDK4) with anti-cancer efficacy in A2780 ovarian cancer culture (IC50 = 760 nM).
1 of 1
This Item | |
|---|---|
| description ≥98% (HPLC) | description ≥98% (HPLC) |
| form powder | form powder |
| assay ≥98% (HPLC) | assay ≥98% (HPLC) |
| Quality Level 100 | Quality Level 100 |
| storage temp. 2-8°C | storage temp. 2-8°C |
| solubility DMSO: 2 mg/mL, clear | solubility DMSO: 2 mg/mL, clear (warmed) |
| color white to beige | color white to beige |
Storage Class
11 - Combustible Solids
WGK
WGK 3
Flash Point (°F)
Not applicable
Flash Point (°C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.

