Skip to Content
MilliporeSigma

Skip To

SML3762

BRD0033

≥98% (HPLC)

Synonym(s):

N-[4-(4-Bromophenyl)-2-thiazolyl]-3-[(4-chlorophenyl)thio]propanamide

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C18H14BrClN2OS2
CAS Number:
Molecular Weight:
453.80
NACRES:
NA.77
UNSPSC Code:
12352200
Assay:
≥98% (HPLC)
Form:
powder
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

-10 to -25°C

Biochem/physiol Actions

BRD0033 is an inactive control for a cell-permeable and potent anti-CRISPR agent BRD7586.
Inactive control for a cell-permeable and potent anti-CRISPR agent BRD7586


Storage Class

11 - Combustible Solids

WGK

WGK 3

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library




Questions

Reviews

No rating value

Active Filters