Skip to Content
MilliporeSigma

Skip To

SML4080

Sakuranetin

≥98% (HPLC)

Synonym(s):

(-)-(S)-Sakuranetin, (2S)-2,3-Dihydro-5-hydroxy-2-(4-hydroxyphenyl)-7-methoxy-4H-1-benzopyran-4-one, (S)-4′,5-Dihydroxy-7-methoxyflavanone, 7-O-Methylnaringenin, Naringenin 7-methyl ether

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C16H14O5
CAS Number:
Molecular Weight:
286.28
MDL number:
Assay:
≥98% (HPLC)
Form:
powder
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

-10 to -25°C

General description

Antifungal phytoalexin; antioxidant, anti-inflammatory, antimycobacterial, antiviral, antifungal, antileishmanial.
Sakuranetin, a 7-O-methylated flavonoid, is an antifungal phytoalexin present in rice that exhibits diverse pharmacological benefits including antioxidant, anti-inflammatory, antimycobacterial, antiviral, antifungal, antileishmanial, antitrypanosomal, glucose uptake stimulation, neuroprotective, antimelanogenic, and antitumor properties.


pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Oral

Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library