Skip to Content
MilliporeSigma

Skip To

SML4099

ABT-639

≥98% (HPLC)

Synonym(s):

(R)-4-Chloro-2-fluoro-N-(2-fluorophenyl)-5-(octahydropyrrolo[1,2-a]pyrazine-2-carbonyl)benzenesulfonamide, 4-Chloro-2-fluoro-N-(2-fluorophenyl)-5-[(8aR)-hexahydropyrrolo[1,2-a]pyrazin-2(1H)-ylcarbonyl]benzenesulfonamide, ABT 639, ABT639

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C20H20ClF2N3O3S
CAS Number:
Molecular Weight:
455.91
MDL number:
NACRES:
NA.21
Assay:
≥98% (HPLC)
Form:
powder
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

-10 to -25°C

SMILES string

Fc1c(cc(c(c1)Cl)C(=O)N3C[C@@H]4N(CC3)CCC4)[S](=O)(=O)Nc2c(cccc2)F

InChI key

AGPIHNZOZNKRGT-CYBMUJFWSA-N

Biochem/physiol Actions

Orally active, peripherally acting, potent and Cav3.2-selective T-type calcium channel blocker with antinociception efficacy in vivo.

ABT-639 is an orally active, peripherally acting, potent and Cav3.2-selective T-type calcium channel blocker (hCav3.2 IC50 = 2 μM vs. >30 μM against Cav3.1/Cav3.3) that attenuates low-voltage-activated (LVA) currents in rat DRG neurons (IC50 = 7.6 μM). ABT-639 exhibits antinociception efficacy in a rat knee joint pain model (ED50 = 2 mg/kg, p.o.), and increases tactile allodynia thresholds in multiple models of neuropathic pain in vivo (10-100 mg/kg, p.o.) with no adverse effects on hemodynamic or psychomotor function even at doses as high as 300 mg/kg.


pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Oral - Skin Irrit. 2

Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library