Skip to Content
MilliporeSigma

Skip To

SML4177

2-NBDG

≥97% (HPLC), powder, fluorescent glucose analog

Synonym(s):

(2R,3R,4S,5R)-3,4,5,6-Tetrahydroxy-2-((7-nitrobenzo[c][1,2,5]oxadiazol-4-yl)amino)hexanal, 2-Deoxy-2-[(7-nitro-2,1,3-benzoxadiazol-4-yl)amino]-D-glucose, 2 NBDG, 2NBDG

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C12H14N4O8
CAS Number:
Molecular Weight:
342.26
MDL number:
NACRES:
NA.21
Assay:
≥97% (HPLC)
Form:
powder
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥97% (HPLC)

form

powder

color

, Faint yellow to dark brown

solubility

H2O: 2 mg/mL, clear (Warmed)

storage temp.

-10 to -25°C

SMILES string

OC[C@@H](O)[C@@H](O)[C@H](O)[C@H](C=O)NC(C1=NON=C12)=CC=C2[N+]([O-])=O

Application

2-Deoxy-2-[(7-nitro-2,1,3-benzoxadiazol-4-yl)-amino]-D-glucose (2-NBDG) may be used as a surrogate reagent to visualize glucose uptake in live cells and tissues for metabolic studies.

Biochem/physiol Actions

2-NBDG, a fluorescent 2-deoxyglucose analog (excitation/emission: ~470/545 nm), supplied as a mixture of α-and β-anomers, enters cells via GLUT transporters, competing with D-glucose. Intracellularly, hexokinase phosphorylates 2-NBDG at C-6, mimicking the first step of glycolysis, before it degrades to a non-fluorescent derivative. This sensitive fluorescent non-toxic tracer illuminates glucose uptake in diverse cell types (e.g., adipocytes, myocytes, tumor cells), offering insights into glycolytic flux, mitochondrial activity, and overall cellular energetics. Compatible with confocal microscopy, flow cytometry, and spectrofluorometry, 2-NBDG (typically used at 10 μM, 37°C, 60 min) enables real-time monitoring of glucose transport kinetics and metabolic dynamics in various biological systems, from insulin-responsive tissues to cancer models.

Features and Benefits

  • offers a balance of ease of use
  • non-toxicity
  • compatibility with common research techniques
  • serves as a valuable tool for studying cellular glucose uptake, utilization, and metabolism in various cell types


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library