Skip to Content
MilliporeSigma

Skip To

SML4250

t-TUCB

≥97% (HPLC), powder, hydrolase (sEH) inhibitor

Synonym(s):

4-[[trans-4-[[[[4-(Trifluoromethoxy)phenyl]amino]carbonyl]amino]cyclohexyl]oxy]benzoic acid, trans-4-[4-[3-(4-Trifluoromethoxyphenyl)ureido]cyclohexyloxy]benzoic acid, EC 1728, EC-1728, EC1728, UC 1728, UC-1728, UC1728

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C21H21F3N2O5
CAS Number:
Molecular Weight:
438.40
MDL number:
NACRES:
NA.21
Assay:
≥97% (HPLC)
Form:
powder
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥97% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

-10 to -25°C

Application

t-TUCB may be used in in vitro studies to examine lipid accumulation and its interaction with omega-3 epoxides in regulating autophagy and endoplasmic reticulum (ER) stress markers.

Biochem/physiol Actions

t-TUCB is an orally active and potent soluble epoxide hydrolase (sEH) inhibitor (h/m/r IC50 = 0.9/1.3/16 nM). t-TUCB modulates inflammation and autophagy in adipose tissue and liver in obese fat-1 mice (10 μg/mL in drinking water, ∼1.67 mg/kg/d). When administered at 10 mg/kg via s.c. t-TUCB is substantially more effective than t-AUCB and APAU in a diabetic neuropathy model in rats.


Storage Class

10 - Combustible liquids

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library