Skip to Content
MilliporeSigma

Skip To

SML4257

Chidamide

≥98% (HPLC), powder, histone deacetylase (HDAC) inhibitor

Synonym(s):

CS 055, CS-055, CS055, HBI 8000, HBI-8000, HBI8000, Tucidinostat, N-(2-Amino-4-fluorophenyl)-4-[[[(2E)-1-oxo-3-(3-pyridinyl)-2-propen-1-yl]amino]methyl]benzamide

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C22H19FN4O2
CAS Number:
Molecular Weight:
390.41
MDL number:
NACRES:
NA.21
Assay:
≥98% (HPLC)
Form:
powder
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

-10 to -25°C

Application

Chidamide may be used as an enhancer of zeste homolog 2 (EZH2) inhibitor and study the antileukemia effects in vivo in leukemia-bearing nude mouse models

Biochem/physiol Actions

Chidamide (CS055, HBI-8000, Tucidinostat) is an orally available benzamide that inhibits against class I histone deaceylases HDAC1/2/3/8 (IC50 of 95/160/67/733 nM), class II2b HDAC10 (IC50 = 78 nM), and class IV HDAC11 (IC50 = 432 nM), but not class IIa HDAC4/5/7/9 or IIb HDAC10 (IC50 >30 μM). Chidamide effectively upregulates cellular acetylation levels of histone H3 and H4 (3 μM for 24 h in SKM-1 and HEL cultures) and exhibits anti-cancer efficacy both in cultures and in vivo.


Pictograms

Exclamation mark

Signal Word

Warning

Hazard Codes

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class

11 - Combustible Solids

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library