Skip to Content
MilliporeSigma

Skip To

SML4270

Martynoside

≥98% (HPLC)

Promotion Available

Synonym(s):

2-(3-Hydroxy-4-methoxyphenyl)ethyl 3-O-(6-deoxy-α-L-mannopyranosyl)-4-O-[(2E)-3-(4-hydroxy-3-methoxyphenyl)-2-propenoyl]-β-D-glucopyranoside, 2-(3-hydroxy-4-methoxyphenyl)ethyl 3-O-(6-deoxy-α-L-mannopyranosyl)- β-D-glucopyranoside 4-[(2E)-3-(4-hydroxy-3-methoxyphenyl)-2-propenoate]

Sign In to View Organizational & Contract Pricing.

Spend $500 — Save 5%; Spend $800 — Save 10%; Spend $1,600 — Save 22%*

Sign in to check eligibility. Shop Products

Select a Size

Change View
Size/SKUAvailabilityPrice
5 mg

Available to ship onSeptember 13, 2026fromMILWAUKEE

$139.00
25 mg

Available to ship onSeptember 13, 2026fromMILWAUKEE

$559.00

About This Item

Empirical Formula (Hill Notation):
C31H40O15
CAS Number:
Molecular Weight:
652.64
Assay:
≥98% (HPLC)
Form:
powder

$139.00

Save Up To 22%


Available to ship onSeptember 13, 2026Details


Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

Biochem/physiol Actions

Martynoside is a bioactive glycoside of Rehmannia glutinosa, a traditional Chinese herb, which exhibits anticancer, cytotoxic, antimetastatic, hematopoietic and chemoprotective activities. Apparently martynosidethe targets ribosomal protein L27a (RPL27A).


Pictograms

Exclamation mark

Signal Word

Warning

Hazard Codes

Hazard Classifications

Skin Sens. 1

Storage Class

11 - Combustible Solids

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library