Skip to Content
MilliporeSigma

Skip To

SML4312

eIF4A3-IN-1

≥97% (HPLC)

Synonym(s):

3-[4-[[(3S)-4-(4-Bromobenzoyl)-3-(4-chlorophenyl)-1-piperazinyl]carbonyl]-5-methyl-1H-pyrazol-1-yl]benzonitrile, Compound 53a

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C29H23BrClN5O2
CAS Number:
Molecular Weight:
588.88
UNSPSC Code:
12352200
Assay:
≥97% (HPLC)
Form:
powder
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥97% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

-10 to -25°C

Biochem/physiol Actions

eIF4A3-IN-1 (Compound 53a) is a highly selective, cell-active inhibitor of eukaryotic initiation factor 4A3 (eIF4A3), exhibiting an ATPase IC50 of 0.26 µM and a KD of 0.043 µM. By directly binding a non-ATP site on eIF4A3, eIF4A3-IN-1 inhibits RNA-dependent ATPase activity while demonstrating excellent selectivity over related helicases eIF4A1, eIF4A2, DHX29, and BRR2 (all IC50 >100 µM). This targeted inhibition subsequently causes dose-dependent cellular suppression of nonsense-mediated RNA decay (NMD), affecting the stability of mRNAs with premature termination codons.


Pictograms

Exclamation mark

Signal Word

Warning

Hazard Codes

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class

11 - Combustible Solids

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library