Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
| Size/SKU | Availability | Price |
|---|
About This Item
Empirical Formula (Hill Notation):
C17H14O5
CAS Number:
Molecular Weight:
298.29
MDL number:
UNSPSC Code:
12352200
Assay:
≥95% (HPLC)
Form:
powder
Pricing and availability is not currently available.
Quality Segment
assay
≥95% (HPLC)
form
powder
color
white to beige
solubility
DMSO: 2 mg/mL, clear
storage temp.
-10 to -25°C
Biochem/physiol Actions
Strigolactone GR24 is potent synthetic agonist of the strigolactone receptor that stimulates and regulates shoot branching. Strigolactone GR24 activates SIRT1 without activating AMPK, enhances insulin signaling, glucose uptake, GLUT4 translocation and mitochondrial biogenesis.
1 of 1
This Item | |||
|---|---|---|---|
| description ≥95% (HPLC) | description - | description - | description - |
| form powder | form - | form - | form - |
| assay ≥95% (HPLC) | assay - | assay - | assay - |
| Quality Level 100 | Quality Level - | Quality Level - | Quality Level - |
| storage temp. -10 to -25°C | storage temp. - | storage temp. - | storage temp. - |
| solubility DMSO: 2 mg/mL, clear | solubility - | solubility - | solubility - |
| color white to beige | color - | color - | color - |
Signal Word
Warning
Hazard Codes
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral
Storage Class
11 - Combustible Solids
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.




