Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
Empirical Formula (Hill Notation):
C26H41BrNO4·Br
CAS Number:
Molecular Weight:
591.42
UNSPSC Code:
12352200
Assay:
≥95% (HPLC)
Form:
powder
Storage condition:
desiccated
Quality Segment
assay
≥95% (HPLC)
form
powder
storage condition
desiccated
color
white to beige
solubility
DMSO: 2 mg/mL, clear
storage temp.
-10 to -25°C
Biochem/physiol Actions
Pinaverium bromide is a calcium channel blocker used as antispasmodics medication for treatment of gastrointestinal disorders. Pinaverium bromide exhibits potent antibacterial activity against Staphylococcus aureus and E. faecalis that rapidly kills S. aureus planktonic cells. Pinaverium bromide potently inhibits growth of S. aureus CHS101 persister cells and biofilm formation.
Disclaimer
Hygroscopic Store with desiccant
Signal Word
Warning
Hazard Codes
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2
Storage Class
11 - Combustible Solids
Flash Point (°F)
Not applicable
Flash Point (°C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
