Skip to Content
MilliporeSigma

Skip To

SML4413

Perfluorohexyloctane

≥98% (GC)

Synonym(s):

1,1,1,2,2,3,3,4,4,5,5,6,6-Tridecafluorotetradecane, 1-(Perfluorohexyl)octane, F6H8, NOV03

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C14H17F13
CAS Number:
Molecular Weight:
432.26
UNSPSC Code:
12352200
Assay:
≥98% (GC)
Form:
oil
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥98% (GC)

form

oil

color

colorless to faint yellow

storage temp.

-10 to -25°C

Biochem/physiol Actions

Perfluorohexyloctane (F6H8) is an ophthalmic medication approved to treat dry eye disease. It appears that perfluorohexyloctane ocular drops decrease corneal surface temperature, which leads to an activation of corneal cold thermoreceptors, followed by an increased reflex lacrimation and blinking.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
446882
description

≥98% (GC)

description

98%

form

oil

form

liquid

assay

≥98% (GC)

assay

98%

Quality Level

100

Quality Level

100

storage temp.

-10 to -25°C

storage temp.

-

color

colorless to faint yellow

color

-


Pictograms

Exclamation mark

Signal Word

Warning

Hazard Codes

Hazard Classifications

Acute Tox. 4 Oral - Skin Irrit. 2

Storage Class

10 - Combustible liquids

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library