Select a Size
| Size/SKU | Availability | Price |
|---|---|---|
20 μg | Usually ships in 2 weeks. | $420.00 |
About This Item
$420.00
recombinant
expressed in baculovirus infected Sf9 cells
assay
≥70% (SDS-PAGE)
form
buffered aqueous glycerol solution
mol wt
~65 kDa
NCBI accession no.
shipped in
dry ice
storage temp.
−70°C
Gene Information
human ... UHMK1(127933)
General description
Physical form
Preparation Note
1 of 1
This Item | |||
|---|---|---|---|
| description recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution | description Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody | description Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution | description IgG fraction of antiserum, buffered aqueous solution |
| recombinant expressed in baculovirus infected Sf9 cells | recombinant - | recombinant - | recombinant - |
| assay ≥70% (SDS-PAGE) | assay - | assay - | assay - |
| form buffered aqueous glycerol solution | form buffered aqueous glycerol solution | form buffered aqueous glycerol solution | form buffered aqueous solution |
| mol wt ~65 kDa | mol wt - | mol wt - | mol wt - |
| shipped in dry ice | shipped in wet ice | shipped in wet ice | shipped in dry ice |
| Gene Information human ... UHMK1(127933) | Gene Information human ... UHMK1(127933) | Gene Information human ... UHMK1(127933) | Gene Information human ... KIS(127933) |
Storage Class
10 - Combustible liquids
WGK
WGK 1
Flash Point (°F)
Not applicable
Flash Point (°C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.



