Skip to Content
MilliporeSigma

Skip To

U5635

UFP-101 trifluoroacetate salt

≥98% (HPLC), powder

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
1 mg
Check Cart for Availability
$304.00

About This Item

Empirical Formula (Hill Notation):
C82H138N32O21 · xC2HF3O2
Molecular Weight:
1908.17 (free base basis)
NACRES:
NA.77
PubChem Substance ID:
UNSPSC Code:
12352200
MDL number:
Assay:
≥98% (HPLC)

$304.00


Check Cart for Availability

​
Request a Custom Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥98% (HPLC)

form

powder

storage temp.

−20°C

SMILES string

OC(=O)C(F)(F)F.C[C@@H](O)[C@H](NC(=O)[C@H](Cc1ccccc1)NC(=O)CNC(=O)CNC(=O)CNCc2ccccc2)C(=O)NCC(=O)N[C@@H](C)C(=O)N[C@@H](CCCNC(N)=N)C(=O)N[C@@H](CCCCN)C(=O)N[C@@H](CO)C(=O)N[C@@H](C)C(=O)N[C@@H](CCCNC(N)=N)C(=O)N[C@@H](CCCCN)C(=O)N[C@@H](CCCNC(N)=N)C(=O)N[C@@H](CCCCN)C(=O)N[C@@H](CC(N)=O)C(=O)N[C@@H](CCC(N)=O)C(N)=O

InChI

1S/C82H138N32O21.C2HF3O2/c1-45(102-64(121)43-101-79(135)66(47(3)116)114-77(133)57(37-48-19-6-4-7-20-48)104-65(122)42-100-63(120)41-99-62(119)40-95-39-49-21-8-5-9-22-49)68(124)106-54(26-16-34-96-80(89)90)71(127)110-53(25-12-15-33-85)75(131)113-59(44-115)78(134)103-46(2)69(125)107-55(27-17-35-97-81(91)92)72(128)108-51(23-10-13-31-83)70(126)111-56(28-18-36-98-82(93)94)73(129)109-52(24-11-14-32-84)74(130)112-58(38-61(87)118)76(132)105-50(67(88)123)29-30-60(86)117;3-2(4,5)1(6)7/h4-9,19-22,45-47,50-59,66,95,115-116H,10-18,23-44,83-85H2,1-3H3,(H2,86,117)(H2,87,118)(H2,88,123)(H,99,119)(H,100,120)(H,101,135)(H,102,121)(H,103,134)(H,104,122)(H,105,132)(H,106,124)(H,107,125)(H,108,128)(H,109,129)(H,110,127)(H,111,126)(H,112,130)(H,113,131)(H,114,133)(H4,89,90,96)(H4,91,92,97)(H4,93,94,98);(H,6,7)/t45-,46-,47+,50-,51-,52-,53-,54-,55-,56-,57-,58-,59-,66-;/m0./s1

InChI key

JBYBTOHYBYFCOD-XLNLJAOCSA-N

Biochem/physiol Actions

UFP-101 is a potent antagonist of nociceptin/orphanin FQ receptor.
UFP-101 is a potent antagonist of nociceptin/orphanin FQ receptor; ORL1


Pictograms

Exclamation mark

Signal Word

Warning

Hazard Codes

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class

11 - Combustible Solids

WGK

WGK 3

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library




Questions

Reviews

No rating value

Active Filters