Skip to Content
MilliporeSigma

Skip To

Z744345

Vacuubrand 2L bottle for BVC system

borosilicate glass, with hose nozzle for connection to the VHCpro

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

UNSPSC Code:
40151700
NACRES:
NB.17
Capacity:
-2 L
Material:
borosilicate glass
Sterility:
non-sterile
Feature:
with cap
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


material

borosilicate glass

sterility

non-sterile

feature

with cap

manufacturer/tradename

Vacuubrand 20635809

capacity

-2 L

General description

2l collection bottle made of borosilicate glass with very high chemical resistance. Plastic shatter protection outside coating to minimize liquid loss in case of glass breakage. Special bottle cap for minimized aerosol and foam formation. Hose nozzle for connection of a VacuuHandControl VHCpro. Additional port to connect a second VHCpro (optional). Filter element 0.2 μm with hydrophobic membrane to protect pump, lines and environment with hose for connecting to a BVC. Completely autoclavable
2L borosilicate glass collection bottle with high chemical resistance, with splinter-proof coating, special bottle head for minimized foam and aerosol formation, hose nozzle for VacuuHandControl VHCproand additional port to connect a second VHCpro (optional), hydrophobic filter element 0.2 µm to protect pump and environment, hose for connection to pump unit BVC or BVC basic; fully autoclavable. 2L borosilicate glass collection bottle with high chemical resistance, with splinter-proof coating, special bottle head for minimized foam and aerosol formation

Application

fluid aspiration accessories

Features and Benefits

  • High chemical resistance
  • Plastic shatter protection outside coating to minimize liquid loss incase of glass breakage
  • Special bottle cap for minimized aerosol and foamformation
  • Hose nozzle for connection of a VacuuHandControl VHCpro with additional port to connect a second VHCpro (optional)
  • Filter element 0.2 μmwith hydrophobic membrane to protect pump, lines and environment withhose for connecting to a BVC
  • Completely autoclavable.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
Z744339Z744346Z744403
description

borosilicate glass, with hose nozzle for connection to the VHCpro

description

for BVC control, BVC professional

description

polypropylene, with hose nozzle for connection to the VHC system

description

2L glass bottle, 100-115V 50/60Hz, 120V 60Hz

feature

with cap

feature

to hold 2L glass bottle in the BVC control or BVC professional pumpunits (included with the complete system for BVC control G and BVCprofessional G)

feature

with cap

feature

Vacuum controller with Touchpanel, Control range 150-850 mbar (Adjustable)

sterility

non-sterile

sterility

-

sterility

non-sterile

sterility

-

capacity

-2 L

capacity

-

capacity

-4 L

capacity

-2 L

material

borosilicate glass

material

plastic

material

polypropylene

material

borosilicate glass bottle

manufacturer/tradename

Vacuubrand 20635809

manufacturer/tradename

Vacuubrand 20635839

manufacturer/tradename

Vacuubrand 20635810

manufacturer/tradename

Vacuubrand 20727503




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library