Skip to Content
MilliporeSigma

Skip To

64101

L-Citrulline-1,2,3,4,5-13C5

certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

product line

TraceCERT®

grade

certified reference material, TraceCERT®

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

format

neat

storage temp.

2-8°C

SMILES string

NC(N[13C][13C][13C][13C](N)[13C](O)=O)=O

General description

This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to the SI through a primary reference material from an NMI, e.g. NIST, NRC or NMIJ.


Certified content incl. uncertainty and expiry date are given on the certificate.


Download your certificate at: http://www.sigma-aldrich.com





Citrulline is a non-proteinogenic amino acid produced in the urea cycle and during nitric oxide synthesis, often used as a biomarker of enterocyte (intestinal) mass and function. Testing citrulline can help assess conditions like short bowel syndrome, mucosal injury from chemotherapy or radiation, and some urea-cycle disorders, supporting both clinical evaluation and research. Accurate measurement commonly uses LC–MS/MS, where a stable isotope–labeled citrulline acts as an internal standard to correct for extraction losses, matrix effects, and instrument variability. Because the labeled standard is chemically identical and co-elutes with native citrulline, it enables robust calibration and improves accuracy, precision, and inter-lab comparability. This leads to more reliable decisions based on citrulline levels.





Discover our portfolio of amino acid reference materals.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
19936748935734187
description

certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

description

certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

description

97 atom % D, 99 atom % 13C, 97% (CP)

description

endotoxin tested, 98 atom % D, 99 atom % 13C, 98% (CP)

Quality Level

300

Quality Level

300

Quality Level

-

Quality Level

-

manufacturer/tradename

Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

manufacturer/tradename

Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

manufacturer/tradename

-

manufacturer/tradename

-

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

-

storage temp.

-

format

neat

format

-

format

-

format

-

shelf life

limited shelf life, expiry date on the label

shelf life

limited shelf life, expiry date on the label

shelf life

-

shelf life

-

product line

TraceCERT®

product line

TraceCERT®

product line

-

product line

-


Storage Class

11 - Combustible Solids

WGK

WGK 3

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library