Skip to Content
MilliporeSigma

Skip To

96646

N-(β-Ketocaproyl-5,5,6,6,6-d5)-DL-homoserine lactone

analytical standard

Synonym(s):

N-(3-Oxohexanoyl-5,5,6,6,6-d5)-DL-homoserine lactone

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C10D5H10NO4
Molecular Weight:
218.26
UNSPSC Code:
12352209
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

analytical standard

Quality Segment

assay

≥98% (HPLC)

shelf life

limited shelf life, expiry date on the label

storage temp.

−20°C


Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
K3255K3007
description

analytical standard

description

analytical standard

description

≥98%

Quality Level

100

Quality Level

300

Quality Level

200

grade

analytical standard

grade

analytical standard

grade

-

assay

≥98% (HPLC)

assay

-

assay

≥98%

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

shelf life

limited shelf life, expiry date on the label

shelf life

-

shelf life

-


Storage Class

11 - Combustible Solids

WGK

WGK 3

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters