Select a Size
| Size/SKU | Availability | Price |
|---|---|---|
100 mg | Check Cart for Availability | $342.00 $342.00 |
About This Item
grade
pharmaceutical primary standard
agency
USP
API family
leucine
form
solid
manufacturer/tradename
USP
mp
187-190 °C (lit.)
application(s)
pharmaceutical (small molecule)
pharmaceutical
pharmaceutical
SMILES string
CC(C)C[C@H](NC(C)=O)C(O)=O
InChI
1S/C8H15NO3/c1-5(2)4-7(8(11)12)9-6(3)10/h5,7H,4H2,1-3H3,(H,9,10)(H,11,12)/t7-/m0/s1
InChI key
WXNXCEHXYPACJF-ZETCQYMHSA-N
General description
Application
Analysis Note
Other Notes
1 of 1
This Item | |||
|---|---|---|---|
| description United States Pharmacopeia (USP) Reference Standard | description ReagentPlus®, 99% | description United States Pharmacopeia (USP) Reference Standard | description United States Pharmacopeia (USP) Reference Standard |
| manufacturer/tradename USP | manufacturer/tradename - | manufacturer/tradename USP | manufacturer/tradename USP |
| grade pharmaceutical primary standard | grade - | grade pharmaceutical primary standard | grade pharmaceutical primary standard |
| form solid | form solid | form - | form - |
| application(s) pharmaceutical (small molecule) | application(s) peptide synthesis | application(s) pharmaceutical (small molecule) | application(s) pharmaceutical (small molecule) |
| API family leucine | API family - | API family sucromalt | API family quinoline yellow |
| mp 187-190 °C (lit.) | mp 187-190 °C (lit.) | mp - | mp - |
Still not finding the right product?
Explore all of our products under N-Acetyl-L-leucine
Storage Class
11 - Combustible Solids
WGK
WGK 3
Flash Point (°F)
Not applicable
Flash Point (°C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.

