Skip to Content
MilliporeSigma

Skip To

1443249

USP

Metyrosine R-Enantiomer

Synonym(s):

(R)-2-Amino-3-(4-hydroxyphenyl)-2-methylpropanoic acid

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C10H13NO3
CAS Number:
Molecular Weight:
195.22
NACRES:
NA.24
UNSPSC Code:
41116107
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

pharmaceutical primary standard

agency

USP

manufacturer/tradename

USP

format

neat

storage temp.

2-30°C

General description

Find answers to frequently asked questions on USP products in USP FAQs.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
144322714432051441288
description

-

description

-

description

United States Pharmacopeia (USP) Reference Standard

description

-

grade

pharmaceutical primary standard

grade

pharmaceutical primary standard

grade

pharmaceutical primary standard

grade

pharmaceutical primary standard

format

neat

format

neat

format

neat

format

neat

storage temp.

2-30°C

storage temp.

2-30°C

storage temp.

-

storage temp.

2-8°C

manufacturer/tradename

USP

manufacturer/tradename

USP

manufacturer/tradename

USP

manufacturer/tradename

USP

agency

USP

agency

USP

agency

USP

agency

USP


Storage Class

11 - Combustible Solids

WGK

WGK 3

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library