Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
| Size/SKU | Availability | Price |
|---|
About This Item
Empirical Formula (Hill Notation):
C27H29ClO6
CAS Number:
Molecular Weight:
484.97
NACRES:
NA.24
UNSPSC Code:
41116107
Pricing and availability is not currently available.
grade
pharmaceutical primary standard
agency
USP
manufacturer/tradename
USP
format
neat
storage temp.
2-30°C
General description
Find answers to frequently asked questions on USP products in USP FAQs.
1 of 1
This Item | |||
|---|---|---|---|
| description - | description United States Pharmacopeia (USP) Reference Standard | description pharmaceutical secondary standard, certified reference material | description certified reference material |
| grade pharmaceutical primary standard | grade pharmaceutical primary standard | grade certified reference material, pharmaceutical secondary standard | grade certified reference material |
| format neat | format neat | format - | format - |
| agency USP | agency USP | agency - | agency - |
| storage temp. 2-30°C | storage temp. - | storage temp. -10 to -25°C | storage temp. -10 to -25°C |
| manufacturer/tradename USP | manufacturer/tradename USP | manufacturer/tradename - | manufacturer/tradename - |
Signal Word
Danger
Hazard Codes
Precautionary Statements
Hazard Classifications
Repr. 1B
Storage Class
6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects
WGK
WGK 3
Flash Point (°F)
Not applicable
Flash Point (°C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.


