Skip to Content
MilliporeSigma

Skip To

1463458

USP

Nilotinib Related Compound D

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

Nilotinib Related Compound D

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C18H16N4O2
CAS Number:
Molecular Weight:
320.35
UNSPSC Code:
41116107
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

pharmaceutical primary standard

agency

USP

manufacturer/tradename

USP

application(s)

pharmaceutical small molecule

format

neat

storage temp.

2-30°C

InChI

1S/C18H16N4O2/c1-12-5-6-13(17(23)24-2)10-16(12)22-18-20-9-7-15(21-18)14-4-3-8-19-11-14/h3-11H,1-2H3,(H,20,21,22)

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including MSDS and any product information leaflets, have been developed and issued under the Authority of the issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  

Other Notes

USP issued SDS can be found here

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
146343914634401463457
description

United States Pharmacopeia (USP) Reference Standard

description

United States Pharmacopeia (USP) Reference Standard

description

United States Pharmacopeia (USP) Reference Standard

description

United States Pharmacopeia (USP) Reference Standard

manufacturer/tradename

USP

manufacturer/tradename

USP

manufacturer/tradename

USP

manufacturer/tradename

USP

grade

pharmaceutical primary standard

grade

pharmaceutical primary standard

grade

pharmaceutical primary standard

grade

pharmaceutical primary standard

format

neat

format

neat

format

neat

format

-

application(s)

pharmaceutical small molecule

application(s)

pharmaceutical (small molecules)

application(s)

pharmaceutical

application(s)

pharmaceutical

agency

USP

agency

USP

agency

USP

agency

USP

storage temp.

2-30°C

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C


Pictograms

Environment

Hazard Codes

Precautionary Statements

Hazard Classifications

Aquatic Chronic 2

Storage Class

11 - Combustible Solids

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library