Skip to Content
MilliporeSigma

Skip To

1800136

USP

1,3-Diazocane-2-thione

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

1,3-Diazocane-2-thione

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
25 mg

Estimated to ship onSeptember 23, 2026fromMILWAUKEE.

$426.00

About This Item

Empirical Formula (Hill Notation):
C6H12N2S
Molecular Weight:
144.24
UNSPSC Code:
41116107

$426.00


Estimated to ship onSeptember 23, 2026. Details


Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

USP Analytical Reference Materials (ARMs)

agency

USP

form

solid

packaging

pkg of 25 mg

manufacturer/tradename

USP

application(s)

pharmaceutical small molecule

format

neat

storage temp.

2-8°C

SMILES string

S=C1NCCCCCN1

InChI

InChI=1S/C6H12N2S/c9-6-7-4-2-1-3-5-8-6/h1-5H2,(H2,7,8,9)

InChI key

OLCIPZHLPOAOIH-UHFFFAOYSA-N

General description

Find answers to frequently asked questions on USP products in USP FAQs.
This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including MSDS and any product information leaflets, have been developed and issued under the Authority of the issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  

Other Notes

Sales restrictions may apply.
USP issued SDS can be found here


Pictograms

Exclamation mark

Signal Word

Warning

Hazard Codes

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class

11 - Combustible Solids

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library