Select a Size
| Size/SKU | Availability | Price |
|---|
About This Item
grade
pharmaceutical analytical impurity (PAI)
agency
USP
form
coarse powder
manufacturer/tradename
USP
application(s)
pharmaceutical small molecule
format
neat
storage temp.
−20°C
SMILES string
O=C(OCCC(C)CCOC(=O)CC[N+]1(C)CCC2=CC(OC)=C(OC)C=C2C1CC3=CC=C(OC)C(OC)=C3)CC[N+]4(C)CCC5=CC(OC)=C(OC)C=C5C4CC6=CC=C(OC)C(OC)=C6.O=S(=O)([O-])C=1C=CC=CC1
InChI
InChI=1S/C54H74N2O12.C6H6O3S/c1-36(20-26-67-53(57)18-24-55(2)22-16-39-32-49(63-8)51(65-10)34-41(39)43(55)28-37-12-14-45(59-4)47(30-37)61-6)21-27-68-54(58)19-25-56(3)23-17-40-33-50(64-9)52(66-11)35-42(40)44(56)29-38-13-15-46(60-5)48(31-38)62-7;7-10(8,9)6-4-2-1-3-5-6/h12-15,30-36,43-44H,16-29H2,1-11H3;1-5H,(H,7,8,9)/q+2;/p-1/t43-,44-,55-,56-;/m1./s1
InChI key
WMADSZWJNPLFCX-MTCXTLHCSA-M
General description
Analysis Note
1 of 1
This Item | |||
|---|---|---|---|
| description Pharmaceutical Analytical Impurity (PAI) | description Pharmaceutical Analytical Impurity (PAI) | description Pharmaceutical Analytical Impurity (PAI) | description Pharmaceutical Analytical Impurity (PAI) |
| grade pharmaceutical analytical impurity (PAI) | grade pharmaceutical analytical impurity (PAI) | grade pharmaceutical analytical impurity (PAI) | grade pharmaceutical analytical impurity (PAI) |
| manufacturer/tradename USP | manufacturer/tradename USP | manufacturer/tradename USP | manufacturer/tradename USP |
| form coarse powder | form powder | form fine powder | form solid |
| format neat | format neat | format neat | format neat |
| application(s) pharmaceutical small molecule | application(s) pharmaceutical small molecule | application(s) pharmaceutical small molecule | application(s) pharmaceutical small molecule |
| storage temp. −20°C | storage temp. −20°C | storage temp. −20°C | storage temp. −20°C |
Signal Word
Warning
Hazard Codes
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral - Skin Irrit. 2
Storage Class
11 - Combustible Solids
Flash Point (°F)
Not applicable
Flash Point (°C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.

