Select a Size
| Size/SKU | Availability | Price |
|---|
About This Item
grade
pharmaceutical analytical impurity (PAI)
agency
USP
manufacturer/tradename
USP
application(s)
pharmaceutical small molecule
format
neat
storage temp.
2-8°C
SMILES string
CN(C)CCC1=CC=C(C=C1)O
InChI
InChI=1S/C10H15NO/c1-11(2)8-7-9-3-5-10(12)6-4-9/h3-6,12H,7-8H2,1-2H3
InChI key
KUBCEEMXQZUPDQ-UHFFFAOYSA-N
General description
Analysis Note
1 of 1
This Item | |||
|---|---|---|---|
| description Pharmaceutical Analytical Impurity (PAI) | description ≥95% (LC/MS-ELSD) | description pharmaceutical analytical impurity (PAI) | description pharmaceutical analytical impurity (PAI) |
| application(s) pharmaceutical small molecule | application(s) metabolomics | application(s) pharmaceutical small molecule | application(s) pharmaceutical small molecule |
| format neat | format - | format neat | format neat |
| grade pharmaceutical analytical impurity (PAI) | grade - | grade pharmaceutical analytical impurity (PAI) | grade pharmaceutical analytical impurity (PAI) |
| storage temp. 2-8°C | storage temp. −20°C | storage temp. - | storage temp. 2-8°C |
| manufacturer/tradename USP | manufacturer/tradename - | manufacturer/tradename USP | manufacturer/tradename USP |
| agency USP | agency - | agency USP | agency USP |
Signal Word
Warning
Hazard Codes
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class
11 - Combustible Solids
Flash Point (°F)
Not applicable
Flash Point (°C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.


