Select a Size
| Size/SKU | Availability | Price |
|---|
About This Item
grade
pharmaceutical analytical impurity (PAI)
agency
USP
form
solid
packaging
pkg of 25 mg
manufacturer/tradename
USP
application(s)
pharmaceutical small molecule
format
neat
storage temp.
2-8°C
SMILES string
Cl.OC1=CC=CC2=C1CCC(N(CC)CCC=3SC=CC3)C2
InChI
InChI=1S/C18H23NOS.ClH/c1-2-19(11-10-16-6-4-12-21-16)15-8-9-17-14(13-15)5-3-7-18(17)20;/h3-7,12,15,20H,2,8-11,13H2,1H3;1H/t15-;/m0./s1
InChI key
CMGIGAZRQHGNIQ-RSAXXLAASA-N
General description
Analysis Note
1 of 1
This Item | |||
|---|---|---|---|
| description Pharmaceutical Analytical Impurity (PAI) | description United States Pharmacopeia (USP) Reference Standard | description United States Pharmacopeia (USP) Reference Standard | description Pharmaceutical Analytical Impurity (PAI) |
| grade pharmaceutical analytical impurity (PAI) | grade pharmaceutical primary standard | grade pharmaceutical primary standard | grade pharmaceutical analytical impurity (PAI) |
| format neat | format neat | format neat | format neat |
| storage temp. 2-8°C | storage temp. 2-8°C | storage temp. - | storage temp. 2-8°C |
| form solid | form - | form - | form solid |
| packaging pkg of 25 mg | packaging - | packaging - | packaging pkg of 25 mg |
| application(s) pharmaceutical small molecule | application(s) pharmaceutical (small molecule) | application(s) pharmaceutical (small molecule) | application(s) pharmaceutical small molecule |
Storage Class
11 - Combustible Solids
Flash Point (°F)
Not applicable
Flash Point (°C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
