Select a Size
| Size/SKU | Availability | Price |
|---|
About This Item
grade
pharmaceutical analytical impurity (PAI)
agency
USP
manufacturer/tradename
USP
application(s)
pharmaceutical small molecule
format
neat
storage temp.
2-8°C
SMILES string
CC(C(=O)C)OC1=CC=C(C=C1)C(=O)C2=CC=C(C=C2)Cl
InChI
InChI=1S/C17H15ClO3/c1-11(19)12(2)21-16-9-5-14(6-10-16)17(20)13-3-7-15(18)8-4-13/h3-10,12H,1-2H3
InChI key
UGIBXSPOZIFSQB-UHFFFAOYSA-N
General description
Analysis Note
1 of 1
This Item | |||
|---|---|---|---|
| description Pharmaceutical Analytical Impurity (PAI) | description pharmaceutical analytical impurity (PAI) | description pharmaceutical analytical impurity (PAI) | description Pharmaceutical Analytical Impurity (PAI) |
| grade pharmaceutical analytical impurity (PAI) | grade pharmaceutical analytical impurity (PAI) | grade pharmaceutical analytical impurity (PAI) | grade pharmaceutical analytical impurity (PAI) |
| format neat | format neat | format neat | format neat |
| agency USP | agency USP | agency USP | agency USP |
| storage temp. 2-8°C | storage temp. 2-8°C | storage temp. 2-8°C | storage temp. 2-8°C |
| application(s) pharmaceutical small molecule | application(s) pharmaceutical small molecule | application(s) pharmaceutical small molecule | application(s) pharmaceutical small molecule |
| manufacturer/tradename USP | manufacturer/tradename USP | manufacturer/tradename USP | manufacturer/tradename USP |
Storage Class
11 - Combustible Solids
Flash Point (°F)
Not applicable
Flash Point (°C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
