Select a Size
| Size/SKU | Availability | Price |
|---|
About This Item
grade
pharmaceutical analytical impurity (PAI)
agency
USP
manufacturer/tradename
USP
application(s)
pharmaceutical small molecule
format
neat
storage temp.
2-8°C
General description
Analysis Note
1 of 1
This Item | |||
|---|---|---|---|
| description pharmaceutical analytical impurity (PAI) | description pharmaceutical analytical impurity (PAI) | description Pharmaceutical Analytical Impurity (PAI) | description pharmaceutical analytical impurity (PAI) |
| manufacturer/tradename USP | manufacturer/tradename USP | manufacturer/tradename USP | manufacturer/tradename USP |
| grade pharmaceutical analytical impurity (PAI) | grade pharmaceutical analytical impurity (PAI) | grade pharmaceutical analytical impurity (PAI) | grade pharmaceutical analytical impurity (PAI) |
| application(s) pharmaceutical small molecule | application(s) pharmaceutical small molecule | application(s) pharmaceutical small molecule | application(s) pharmaceutical small molecule |
| format neat | format neat | format neat | format neat |
| storage temp. 2-8°C | storage temp. - | storage temp. 2-8°C | storage temp. 2-8°C |
| agency USP | agency USP | agency USP | agency USP |
Still not finding the right product?
Explore all of our products under Baccatin III
Signal Word
Danger
Hazard Codes
Precautionary Statements
Hazard Classifications
Carc. 1B - Eye Irrit. 2 - Muta. 1B - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class
6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects
Flash Point (°F)
Not applicable
Flash Point (°C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.


