MilliporeSigma
Tangential Flow Filtration Devices
Our tangential flow filtration (TFF) products provide versatile solutions for both single-use and multi-use pharma and biopharma applications. Designed to streamline processes, these devices maximize yield and product recovery, enhancing efficiency while minimizing operational complexity.
Our Pellicon® Capsules and cassettes can be used for different applications and offer options for every ultrafiltration and diafiltration challenge.
Pellicon® single-pass TFF is a convenient way to reduce volumes to eliminate tank bottlenecks, reduce the size of subsequent steps, concentrate with inline buffer addition for desalting, and facilitate high final concentrations with high product recovery
SINGLE-PASS TFF WITH PELLICON® CAPSULES AND CASSETTES 
Single-pass TFF with our Pellicon® capsules and cassettes streamlines processing by concentrating process streams without the recirculation of traditional TFF. This is a simple way to increase capacity, concentration, and product recovery while improving footprint. This versatile application also enables continuous and intensified processing, in which in-line volume reduction is coupled to other process steps.
Our single-use Pellicon® Capsules can be ready to perform ultrafiltration and diafiltration of mAbs and ADCs in minutes
Pellicon® Capsules
Pellicon® Capsules with Ultracel® membrane combine streamlined ultrafiltration and diafiltration with ease of use and process flexibility. This innovative single-use TFF capsule delivers superior mass transfer, flux and optimum recovery in a unique plug-and-play, holderless device that comes gamma sterilized and ready to process in minutes.
Pellicon® 3 cassettes deliver high protein yield with consistent purity and quality for a variety of drug products
Pellicon® 3 Cassettes
Pellicon® 3 cassettes deliver high protein yield and consistent performance while offering broad chemical compatibility. They are available with Ultracel® and Biomax® membranes for different processing needs, including mAbs, plasma, insulin, peptides, and vaccines. To overcome the filtration challenges of high viscosity, our Pellicon® 3 cassettes with D screen are easy to use, enable faster processing and are optimized to achieve higher protein concentrations.
Explore capabilities of Pellicon® 3 Cassettes in this Performance Guide. 
Pellicon® 2 Cassettes
Pellicon® 2 cassettes are ideal for the ultrafiltration and diafiltration of biologics including concentration, desalting, and buffer exchange of polysaccharides, virus suspensions, and protein solutions. Our Ultracel® and Biomax® membranes offer efficient ultrafiltration while Durapore® membrane is best suited for microfiltration.
Pellicon® XL 50 Cassettes
Pellicon® XL 50 cassettes couple our ultrafiltration and microfiltration membranes with linear scalability for process development or small-volume processing.
Prostak™ Microfiltration Modules
Prostak™ microfiltration modules provide robust construction, reliable performance, and steam-in-place capability, making them suitable for clarification, cell harvest, and multi-use perfusion. The low hold-up volume and open channel design allow high product recovery and gentle flow path for processing shear-sensitive materials.
Support for Qualification, Risk Assessment, and Process Optimization
Complementing our product portfolio, the Emprove® Program provides three types of dossiers to support different stages of development and manufacturing operations such as qualification, risk assessment, and process optimization. The dossiers consolidate comprehensive product-specific testing data, quality statements, and regulatory information in a readily available format to simplify your compliance needs.
Products
Sort by Relevance
Showing 1-20 of 241
Change View
Showing 1-20 of 241
Compare
Product Number
Product Name
Product Description
Pricing
Related Articles
Related Resources
Page 1 of 4