Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
| Size/SKU | Availability | Price |
|---|---|---|
50 mL | Check Cart for Availability | $237.00 |
250 mL | Check Cart for Availability | $738.00 |
About This Item
NACRES:
NA.77
UNSPSC Code:
41116012
$237.00
Check Cart for Availability
form
liquid
Quality Segment
feature
for stabilization of DNA, RNA, and protein in tissue extracts
input
general use sample(s)
shipped in
wet ice
storage temp.
2-8°C
General description
The stability of protein and antibody structure and function is dependent on a variety of factors such as temperature, freeze/thaw cycles, proteases and pH. For some proteins and antibodies, function can be lost completely with a single freeze/thaw cycle and proteases can significantly degrade proteins and antibodies within hours even when kept at 4 °C. Stabilyser Reagent minimizes the risk to improve protein assay quality and accuracy.
Stabilyser Reagent is a proprietary formulation of detergents and solutes optimized for efficient extraction, stabilization and storage of protein, DNA, and RNA from tissue samples. The unique composition of Stabilyser enables isolation of functionally active proteins, while maintaining integrity of nucleic acids. Stabilization of proteins and RNA is achieved even after 24 hours at room temperature Stabilyser reagent is compatible with Western blotting protocols, functional protein activity assays, and nucleic acid applications like qPCR.
Use in conjunction with a homognenizer for simultaneous tissue lysis and stabilization of protein and nucleic acids in tissue samples.
Stabilyser Reagent is a proprietary formulation of detergents and solutes optimized for efficient extraction, stabilization and storage of protein, DNA, and RNA from tissue samples. The unique composition of Stabilyser enables isolation of functionally active proteins, while maintaining integrity of nucleic acids. Stabilization of proteins and RNA is achieved even after 24 hours at room temperature Stabilyser reagent is compatible with Western blotting protocols, functional protein activity assays, and nucleic acid applications like qPCR.
Use in conjunction with a homognenizer for simultaneous tissue lysis and stabilization of protein and nucleic acids in tissue samples.
Application
Prevents degradation of proteins and nucleic acid during tissue lysis and storage. Stabilize functional proteins and RNA.
Features and Benefits
- Gentle, non-denaturing lysis buffer does not interfere with downstream applications
- Convenient ready-to-use reagent
- Stabilizes RNA and protein for analysis at a later date
Preparation Note
Ready-to-use reagent. In general it is recommended to start optimization with 20 μL of Stabilyser reagent per mg of tissue.
1 of 1
This Item | |||
|---|---|---|---|
| description for stabilization of DNA, RNA, and protein in tissue extracts | description Stabilize and protect RNA with immediate RNase inactivation | description LS, For processing fluid samples such as cell suspensions, CSF, and amniotic fluid. | description - |
| storage temp. 2-8°C | storage temp. - | storage temp. - | storage temp. - |
| input general use sample(s) | input - | input - | input - |
| shipped in wet ice | shipped in - | shipped in - | shipped in - |
| Quality Level 200 | Quality Level 100 | Quality Level 100 | Quality Level 100 |
| form liquid | form - | form - | form liquid |
| feature for stabilization of DNA, RNA, and protein in tissue extracts | feature - | feature - | feature - |
Storage Class
10 - Combustible liquids
WGK
WGK 2
Flash Point (°F)
Not applicable
Flash Point (°C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.