Milli-Q® CLX 7000 Connected High-Throughput Water Purification Systems for Clinical Analyzers
The most advanced water purification systems to support maximum lab productivity
Milli-Q® CLX 7000 connected purification systems provide an economical and reliable high-performance water purification solution for clinical analyzers with pure water needs up to 3000 L daily. Water produced complies with the Clinical and Laboratory Standards Institute® (CLSI) guideline for clinical laboratory reagent water (CLRW).
These high-throughput biomedical water systems are designed to maximize your lab’s productivity by integrating our state-of-the-art purification technologies and delivering an unprecedented level of system accessibility and service.
Features & Benefits
  • Delivers consistent CLRW quality water that exceeds CLSI® standards. This means accurate and reproducible diagnostic test results.
  • Incorporates the most advanced water purification technologies, including patented Elix® electrodeionization (EDI) module and E.R.A.™ (Evolutive Reject Adjustment) technology. Biomedical lab users can focus on their work, save time, and reduce running costs.
  • New Milli-Q® Connect remote service and monitoring capability is now available on all Milli-Q® CLX 7000 systems. This secure online portal simplifies system management and supports labs in achieving maximum productivity.
  • Milli-Q® Connect supports full data traceability, effortless data access and retrieval, and facilitates audit preparation and lab accreditation
  • User-friendly touchscreen makes the system easy to use, with all system information and water quality data rapidly accessible.
  • Quiet, compact and mobile. Install this water system wherever it’s convenient in the lab.
Water Applications
Biomedical Applications
  • Chemistry, Immunochemistry
  • Automation
  • Hematology
  • Microbiology
  • Histology
  • Molecular biology
Water Specifications
Milli-Q® CLX 7000 Series (Milli-Q® CLX 7040 / Milli-Q® CLX 7080 / Milli-Q® CLX 7120 / Milli-Q® CLX 7150)
Resistivity at 25°C
> 15  MΩ.cm
TOC (Total Organic Carbon) Typically < 30 ppb
Bacteria Typically < 1 cfu/mL
Dissolved silica < 0.05 mg/L
Production flow rate 40 L/h up to 150 L/h
Distribution flow rate to analyzers 4 L/min
Distribution pressure to analyzers Adjustable : 0.9 up to 2.1 bar*
* With a distribution loop length of 20 m.
Video – Milli-Q® Connect Remote Services
Supports maximum lab productivity. Now available on all Milli-Q® CLX 7000 systems.