Merck
  • Genetic variation influences immune responses in sensitive rats following exposure to TiO2 nanoparticles.
Genetic variation influences immune responses in sensitive rats following exposure to TiO2 nanoparticles.
Toxicology (2014-12-03)
Asa Gustafsson, Sofia Jonasson, Thomas Sandström, Johnny C Lorentzen, Anders Bucht
ZUSAMMENFASSUNG
This study examines the immunological responses in rats following inhalation to titanium dioxide nanoparticles (TiO2 NPs), in naïve rats and in rats with induced allergic airway disease. The responses of two different inbred rat strains were compared: the Dark Aguoti (DA), susceptible to chronic inflammatory disorders, and the Brown Norwegian (BN), susceptible to atopic allergic inflammation. Naïve rats were exposed to an aerosol of TiO2 NPs once daily for 10 days. Another subset of rats was sensitized to the allergen ovalbumin (OVA) in order to induce airway inflammation. These sensitized rats were exposed to TiO2 NPs before and during the allergen challenge. Naïve rats exposed to TiO2 NPs developed an increase of neutrophils and lymphocytes in both rat strains. Airway hyperreactivity and production of inflammatory mediators typical of a T helper 1 type immune response were significantly increased, only in DA rats. Sensitization of the rats induced a prominent OVA-specific-IgE and IgG response in the BN rat while DA rats only showed an increased IgG response. Sensitized rats of both strains developed airway eosinophilia following allergen challenge, which declined upon exposure to TiO2 NPs. The level of neutrophils and lymphocytes increased upon exposure to TiO2 NPs in the airways of DA rats but remained unchanged in the airways of BN rats. In conclusion, the responses to TiO2 NPs were strain-dependent, indicating that genetics play a role in both immune and airway reactivity. DA rats were found to be higher responder compared to BN rats, both when it comes to responses in naïve and sensitized rats. The impact of genetically determined factors influencing the inflammatory reactions pinpoints the complexity of assessing health risks associated with nanoparticle exposures.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Albumin aus Hühnereiweiss, lyophilized powder, ≥98% (agarose gel electrophoresis)
Sigma-Aldrich
Titan(IV)-oxid, nanopowder, 21 nm primary particle size (TEM), ≥99.5% trace metals basis
Sigma-Aldrich
Titan(IV)-oxid, Anatase, nanopowder, <25 nm particle size, 99.7% trace metals basis
Sigma-Aldrich
3,3′,5,5′-Tetramethylbenzidin, ≥99%
Sigma-Aldrich
Titan(IV)-oxid, Anatase, powder, 99.8% trace metals basis
Sigma-Aldrich
Titan(IV)-oxid, ReagentPlus®, ≥99%
Sigma-Aldrich
Titan(IV)-oxid, Anatase, powder, −325 mesh, ≥99% trace metals basis
Sigma-Aldrich
Albumin aus Hühnereiweiss, lyophilized powder, ≥98% (agarose gel electrophoresis)
Sigma-Aldrich
Titan(IV)-oxid, Rutil, powder, <5 μm, ≥99.9% trace metals basis
Sigma-Aldrich
Titan(IV)-oxid, Rutil, nanopowder, <100 nm particle size, 99.5% trace metals basis
Sigma-Aldrich
3,3′,5,5′-Tetramethylbenzidin, ≥98% (TLC)
Sigma-Aldrich
Albumin aus Hühnereiweiss, powder, 62-88% (agarose gel electrophoresis)
Sigma-Aldrich
Titan(IV)-oxid, Mischung aus Rutil und Anatas, nanopowder, <100 nm particle size (BET), 99.5% trace metals basis
Sigma-Aldrich
Titan, foil, thickness 0.127 mm, 99.7% trace metals basis
Sigma-Aldrich
Albumin aus Hühnereiweiss, lyophilized powder
Sigma-Aldrich
3,3′,5,5′-Tetramethylbenzidin, ≥98.0% (NT)
Sigma-Aldrich
Albumin aus Hühnereiweiss, lyophilized powder, ≥90% (agarose gel electrophoresis)
Sigma-Aldrich
Titan, foil, thickness 0.25 mm, 99.7% trace metals basis
Sigma-Aldrich
Titan, powder, <45 μm avg. part. size, 99.98% trace metals basis
Sigma-Aldrich
Titan, powder, −100 mesh, 99.7% trace metals basis
Sigma-Aldrich
3,3′,5,5′-Tetramethylbenzidin, tablet, 1 mg substrate per tablet
Sigma-Aldrich
Titan(IV)-oxid, Mischung aus Rutil und Anatas, nanoparticles, <150 nm particle size (volume distribution, DLS), dispersion, 40 wt. % in H2O, 99.5% trace metals basis
Sigma-Aldrich
Titan(IV)-oxid, Rutil, ≥99.98% trace metals basis
Supelco
Albumin aus Hühnereiweiss, For use as a marker in SDS-PAGE
Sigma-Aldrich
Titan(IV)-oxid, Rutil, 99.995% trace metals basis
Titan, mesh, 100x100mm, nominal aperture 0.19mm, wire diameter 0.23mm, 60x60 wires/inch, open area 20%, twill weave
Sigma-Aldrich
Titan(IV)-oxid, contains 1% Mn as dopant, nanopowder, <100 nm particle size (BET), ≥97%
Sigma-Aldrich
Titan, wire, diam. 0.25 mm, 99.7% trace metals basis
Sigma-Aldrich
Titan, sponge, 1-20 mm, 99.5% trace metals basis
Sigma-Aldrich
Titan, sputtering target, diam. × thickness 2.00 in. × 0.25 in., 99.995% trace metals basis