Merck
  • Characterization and optimization of heroin hapten-BSA conjugates: method development for the synthesis of reproducible hapten-based vaccines.
Characterization and optimization of heroin hapten-BSA conjugates: method development for the synthesis of reproducible hapten-based vaccines.
Analytical and bioanalytical chemistry (2014-08-03)
Oscar B Torres, Rashmi Jalah, Kenner C Rice, Fuying Li, Joshua F G Antoline, Malliga R Iyer, Arthur E Jacobson, Mohamed Nazim Boutaghou, Carl R Alving, Gary R Matyas
ZUSAMMENFASSUNG
A potential new treatment for drug addiction is immunization with vaccines that induce antibodies that can abrogate the addictive effects of the drug of abuse. One of the challenges in the development of a vaccine against drugs of abuse is the availability of an optimum procedure that gives reproducible and high yielding hapten-protein conjugates. In this study, a heroin/morphine surrogate hapten (MorHap) was coupled to bovine serum albumin (BSA) using maleimide-thiol chemistry. MorHap-BSA conjugates with 3, 5, 10, 15, 22, 28, and 34 haptens were obtained using different linker and hapten ratios. Using this optimized procedure, MorHap-BSA conjugates were synthesized with highly reproducible results and in high yields. The number of haptens attached to BSA was compared by 2,4,6-trinitrobenzenesulfonic acid (TNBS) assay, modified Ellman's test and matrix assisted laser desorption ionization time-of-flight mass spectrometry (MALDI-TOF MS). Among the three methods, MALDI-TOF MS discriminated subtle differences in hapten density. The effect of hapten density on enzyme-linked immunosorbent assay (ELISA) performance was evaluated with seven MorHap-BSA conjugates of varying hapten densities, which were used as coating antigens. The highest antibody binding was obtained with MorHap-BSA conjugates containing 3-5 haptens. This is the first report that rigorously analyzes, optimizes and characterizes the conjugation of haptens to proteins that can be used for vaccines against drugs of abuse. The effect of hapten density on the ELISA detection of antibodies against haptens demonstrates the importance of careful characterization of the hapten density by the analytical techniques described.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Acetonitril, suitable for HPLC, gradient grade, ≥99.9%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Dimethylsulfoxid, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Dimethylsulfoxid, ACS reagent, ≥99.9%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Trifluoressigsäure, ReagentPlus®, 99%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Dimethylsulfoxid, Molecular Biology
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Dimethylsulfoxid, suitable for HPLC, ≥99.7%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Trifluoressigsäure, suitable for HPLC, ≥99.0%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Kaliumphosphat, ACS reagent, ≥99.0%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Acetonitril, HPLC Plus, ≥99.9%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Dimethylsulfoxid, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Ameisensäure, reagent grade, ≥95%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Dimethylsulfoxid, ReagentPlus®, ≥99.5%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Dimethylsulfoxid, anhydrous, ≥99.9%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Natriumdodecylsulfat, BioReagent, suitable for electrophoresis, Molecular Biology, ≥98.5% (GC)
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Acetonitril, ACS reagent, ≥99.5%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Ameisensäure, ACS reagent, ≥96%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Acetonitril, anhydrous, 99.8%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Trifluoressigsäure, puriss. p.a., suitable for HPLC, ≥99.0% (GC)
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Dimethylsulfoxid, ≥99.5% (GC), suitable for plant cell culture
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Natriumdodecylsulfat, ≥99.0% (GC), dust-free pellets
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Natriumchlorid, Molecular Biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Kaliumphosphat, powder, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.0%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Picrylsulfonsäure -Lösung, 5 % (w/v) in H2O, BioReagent, suitable for determination of primary amines
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Natriumchlorid -Lösung, 5 M in H2O, BioReagent, Molecular Biology, suitable for cell culture
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Acetonitril, suitable for HPLC, gradient grade, ≥99.9%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Natriumchlorid, BioXtra, ≥99.5% (AT)
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Natriumchlorid, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Natriumchlorid -Lösung, 0.9% in water, BioXtra, suitable for cell culture
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Natriumdodecylsulfat, ACS reagent, ≥99.0%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Natriumdodecylsulfat -Lösung, BioUltra, Molecular Biology, 10% in H2O
Preise und Verfügbarkeit sind derzeit nicht verfügbar.