Merck
  • New topical photodynamic therapy for treatment of hidradenitis suppurativa using methylene blue niosomal gel: a single-blind, randomized, comparative study.
New topical photodynamic therapy for treatment of hidradenitis suppurativa using methylene blue niosomal gel: a single-blind, randomized, comparative study.
Clinical and experimental dermatology (2014-09-30)
M A Fadel, A A Tawfik
ZUSAMMENFASSUNG
Hidradenitis suppurativa (HS), is a chronic, recurrent dermatosis affecting skin that contains apocrine glands. Photodynamic therapy using aminolaevulinic acid (ALA) activated by intense pulsed light (IPL) have shown variable success rates, with some adverse effects. To evaluate the efficacy and safety of methylene blue (MB) as a photosensitizer delivered as a niosomal gel for the treatment of HS using IPL. We enrolled 11 patients with HS in the study, which was a randomized split-body study. One side of each patient's body was treated with niosomal MB (NMB) gel and the other side was treated with unloaded (free) MB (FMB) gel. The affected sites were irradiated using IPL with a 630 nm filter. Patients were followed up at 1, 3 and 6 months after treatment. Drug release from the FMB gel was significantly higher (P > 0.05) than from the NMB gel. Lesions showed 77.3% and 44.1% reduction on the NMB and FMB sides, respectively. A significant reduction in the Hidradenitis Suppurativa Lesion, Area and Severity Index (HS-LASI) after treatment was elicited in both groups, with no pain, erythema or hyperpigmentation. The combination of MB as a photosensitizer activated with 630 nm IPL as a light source is a successful PDT for HS. Delivery of MB in niosomes was more effective for drug penetration to the dermis compared with delivery by FMB gel. The 630 nm filter was not only a source of activation of MB but also a means of hair-follicle destruction.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Chloroform, contains 100-200 ppm amylenes as stabilizer, ≥99.5%
Sigma-Aldrich
Chloroform, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%, contains amylenes as stabilizer
Sigma-Aldrich
Chloroform, contains ethanol as stabilizer, ACS reagent, ≥99.8%
Sigma-Aldrich
Chloroform, suitable for HPLC, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Cholesterin, Sigma Grade, ≥99%
Supelco
Methylparaben, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Chloroform, contains amylenes as stabilizer, ACS reagent, ≥99.8%
Sigma-Aldrich
Chloroform, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Cholesterin, powder, BioReagent, suitable for cell culture, ≥99%
Sigma-Aldrich
Chloroform, ACS spectrophotometric grade, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, ReagentPlus®, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Methyl-4-hydroxybenzoat, ReagentPlus®, ≥99.0%, crystalline
Sigma-Aldrich
Chloroform, anhydrous, ≥99%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, anhydrous, contains amylenes as stabilizer, ≥99%
Sigma-Aldrich
SyntheChol® NS0-Supplement, 500 ×, synthetic cholesterol, animal component-free, sterile-filtered, aqueous solution, suitable for cell culture
Sigma-Aldrich
Chloroform, contains ethanol as stabilizer, meets analytical specification of BP, 99-99.4% (GC)
USP
Methylparaben, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Cholesterin, from sheep wool, ≥92.5% (GC), powder
Sigma-Aldrich
Chloroform, biotech. grade, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Supelco
Cholesterin, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Methylparaben, PESTANAL®, analytical standard
Supelco
Cholesterin -Lösung, certified reference material, 10 mg/mL in chloroform
Sigma-Aldrich
Methyl-4-hydroxybenzoat, BioReagent, suitable for insect cell culture
Supelco
Chloroform, analytical standard
Sigma-Aldrich
Methyl-4-hydroxybenzoat, ≥99%, FCC
Supelco
Chloroform, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Chloroform, ≥99%, PCR Reagent, contains amylenes as stabilizer
Methylparahydroxybenzoat, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Sorbitanmonostearat, meets FCC analytical specifications
Supelco
Chloroform, Pharmaceutical Secondary Standard; Certified Reference Material