Merck
  • Application of optimized dispersive liquid-liquid microextraction for determination of melatonin by HPLC-UV in plasma samples.
Application of optimized dispersive liquid-liquid microextraction for determination of melatonin by HPLC-UV in plasma samples.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences (2014-05-02)
M S Talebianpoor, S Khodadoust, A Rozbehi, M Akbartabar Toori, M Zoladl, M Ghaedi, R Mohammadi, A S Hosseinzadeh
ZUSAMMENFASSUNG
Melatonin (N-acetyl-3-(2-aminoethyl)-5-methoxyindole) is biologically active as a neurohormone and antioxidant agent. The optimized dispersive liquid-liquid microextraction (DLLME) followed by high-performance liquid chromatography-ultra violet detection (HPLC-UV) was used for the analysis of melatonin in human plasma. Influence variables such as volume of extracting (carbon tetrachloride: CCl4) and dispersing solvents (acetonitrile: ACN), pH and ionic strength, extraction time and centrifugation time were screened in a 2(6-2) fractional factorial design (FFD) and then the significant variables were optimized by using a central composite design (CCD). At optimum conditions values of variables set as pH 6.0, 1.5 mL ACN, 140 μL CCl4, 1.0 min extraction time and 3.0 min centrifugation at 4,500 rpm. At optimum conditions method has linear response over 2.0-500.0 ng mL(-1) with detection limit of 0.5 ng mL(-1) with relative standard deviations (RSDs) less than 5.0%. The values of intra-day and inter-day RSD were 4.3% and 8.5%, respectively. The method was applied successfully for the analysis of melatonin in plasma sample.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Acetonitril, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Ethyl alcohol, Pure 200 proof, Molecular Biology
Sigma-Aldrich
Aceton, ACS reagent, ≥99.5%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, ACS reagent, ≥99.5%
Sigma-Aldrich
Dichlormethan, suitable for HPLC, ≥99.8%, contains amylene as stabilizer
Sigma-Aldrich
Aceton, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Acetonitril, HPLC Plus, ≥99.9%
Sigma-Aldrich
Dichlormethan, contains 40-150 ppm amylene as stabilizer, ACS reagent, ≥99.5%
Sigma-Aldrich
Chloroform, contains 100-200 ppm amylenes as stabilizer, ≥99.5%
Sigma-Aldrich
Aceton, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%
Sigma-Aldrich
Dichlormethan, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%, contains 50-150 ppm amylene as stabilizer
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Chloroform, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%, contains amylenes as stabilizer
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, meets USP testing specifications
Sigma-Aldrich
Acetonitril, ACS reagent, ≥99.5%
Sigma-Aldrich
Chloroform, contains ethanol as stabilizer, ACS reagent, ≥99.8%
Sigma-Aldrich
Chloroform, suitable for HPLC, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Dichlormethan, anhydrous, ≥99.8%, contains 40-150 ppm amylene as stabilizer
Sigma-Aldrich
Acetonitril, anhydrous, 99.8%
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Natriumchlorid, Molecular Biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Ethyl alcohol, Pure 190 proof, for molecular biology
Sigma-Aldrich
Natriumchlorid -Lösung, 5 M in H2O, BioReagent, Molecular Biology, suitable for cell culture
Sigma-Aldrich
Acetonitril, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Natriumchlorid, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Natriumchlorid, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Natriumchlorid -Lösung, 0.9% in water, BioXtra, suitable for cell culture