Merck
  • The effects of cigarette smoke extract on ovulation, oocyte morphology and ovarian gene expression in mice.
The effects of cigarette smoke extract on ovulation, oocyte morphology and ovarian gene expression in mice.
PloS one (2014-04-30)
Zixin Mai, Ming Lei, Bolan Yu, Hongzi Du, Jianqiao Liu
ZUSAMMENFASSUNG
Cigarette smoking can harm fertility, but the existing research has targeted primarily on ovarian follicles, embryos or sex hormone. In this study, we tested cigarette smoke extract on ovulation, oocyte morphology and ovarian gene expression associated with inhibition of oxidative stress using C57BL/6 mice. Mice in the experimental group were administered a cigarette smoke extract (CSE) solution (2 mg/ml) orally daily, while the blank control group was given dimethylsulfoxide (DMSO). A positive control group (menadione) was used that received an intraperitoneal injection of 15 mg/kg menadione in oil solution daily. We found that the CSE group manifested a reduced diameter of zona pellucida-free oocyte (ZP-free OD) and a morphologically misshapen first polar body (PB). Our results suggest that CSE exposure is associated with a shrink size and poor quality of oocytes. Quitting smoking is a wise choice to ensure good fertility.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Dimethylsulfoxid, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethylsulfoxid, ACS reagent, ≥99.9%
Sigma-Aldrich
Dimethylsulfoxid, Molecular Biology
Sigma-Aldrich
Dimethylsulfoxid, suitable for HPLC, ≥99.7%
Sigma-Aldrich
Dimethylsulfoxid, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dimethylsulfoxid, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Dimethylsulfoxid, anhydrous, ≥99.9%
Sigma-Aldrich
Dimethylsulfoxid, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Magnesiumchlorid -Lösung, Molecular Biology, 1.00 M±0.01 M
Sigma-Aldrich
Magnesiumchlorid, ≥98%
Sigma-Aldrich
Dimethylsulfoxid, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Magnesiumchlorid, powder, <200 μm
Sigma-Aldrich
Dimethylsulfoxid, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Menadion, crystalline
Sigma-Aldrich
Magnesiumchlorid -Lösung, BioUltra, Molecular Biology, 2 M in H2O
Sigma-Aldrich
Magnesiumchlorid -Lösung, BioUltra, Molecular Biology, ~1 M in H2O
Sigma-Aldrich
Dimethylsulfoxid, PCR Reagent
Sigma-Aldrich
Magnesiumchlorid, suitable for insect cell culture, BioReagent, ≥97.0%
Sigma-Aldrich
Magnesiumchlorid -Lösung, PCR Reagent, 25 mM MgCI2 solution for PCR
Sigma-Aldrich
Menadion, meets USP testing specifications
Supelco
Menadion (K3), analytical standard
Sigma-Aldrich
Magnesiumchlorid, AnhydroBeads, −10 mesh, 99.9% trace metals basis
Supelco
Dimethylsulfoxid, analytical standard
Sigma-Aldrich
Magnesiumchlorid, AnhydroBeads, −10 mesh, 99.99% trace metals basis
Supelco
Dimethylsulfoxid, for inorganic trace analysis, ≥99.99995% (metals basis)
USP
Dimethylsulfoxid, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Dimethylsulfoxid -Lösung, 50 wt. % in H2O
Supelco
Menadion, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Magnesiumchlorid -Lösung, 0.1 M
Dimethylsulfoxid, European Pharmacopoeia (EP) Reference Standard