Merck
  • Liquid chromatography tandem mass spectrometry determination of selected synthetic cathinones and two piperazines in oral fluid. Cross reactivity study with an on-site immunoassay device.
Liquid chromatography tandem mass spectrometry determination of selected synthetic cathinones and two piperazines in oral fluid. Cross reactivity study with an on-site immunoassay device.
Journal of chromatography. A (2014-12-09)
Ana de Castro, Elena Lendoiro, Hadriana Fernández-Vega, Stefan Steinmeyer, Manuel López-Rivadulla, Angelines Cruz
ZUSAMMENFASSUNG
Since the past few years, several synthetic cathinones and piperazines have been introduced into the drug market to substitute illegal stimulant drugs such as amphetamine and derivatives or cocaine due to their unregulated situation. These emerging drugs are not usually included in routine toxicological analysis. We developed and validated a LC-MS/MS method for the determination of methedrone, methylone, mephedrone, 3,4-methylenedioxypyrovalerone (MDPV), fluoromethcathinone, fluoromethamphetamine, 1-(3-chlorophenyl)piperazine (mCPP) and 3-trifluoromethylphenylpiperazine (TFMPP) in oral fluid. Sample extraction was performed using Strata X cartridges. Chromatographic separation was achieved in 10min using an Atlantis(®) T3 column (100mm×2.1mm, 3μm), and formic acid 0.1% and acetonitrile as mobile phase. The method was satisfactorily validated, including selectivity, linearity (0.2-0.5 to 200ng/mL), limits of detection (0.025-0.1ng/mL) and quantification (0.2-0.5ng/mL), imprecision and accuracy in neat oral fluid (%CV=0.0-12.7% and 84.8-103.6% of target concentration, respectively) and in oral fluid mixed with Quantisal™ buffer (%CV=7.2-10.3% and 80.2-106.5% of target concentration, respectively), matrix effect in neat oral fluid (-11.6 to 399.7%) and in oral fluid with Quantisal™ buffer (-69.9 to 131.2%), extraction recovery (87.9-134.3%) and recovery from the Quantisal™ (79.6-107.7%), dilution integrity (75-99% of target concentration) and stability at different conditions (-14.8 to 30.8% loss). In addition, cross reactivity produced by the studied synthetic cathinones in oral fluid using the Dräger DrugTest 5000 was assessed. All the analytes produced a methamphetamine positive result at high concentrations (100 or 10μg/mL), and fluoromethamphetamine also at low concentration (0.075μg/mL).
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Acetonitril, suitable for HPLC, gradient grade, ≥99.9%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
2-Propanol, suitable for HPLC, 99.9%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
2-Propanol, ACS reagent, ≥99.5%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Dichlormethan, suitable for HPLC, ≥99.8%, contains amylene as stabilizer
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Acetonitril, HPLC Plus, ≥99.9%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Dichlormethan, contains 40-150 ppm amylene as stabilizer, ACS reagent, ≥99.5%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Kaliumchlorid, ACS reagent, 99.0-100.5%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Natriumhydroxid, ACS reagent, ≥97.0%, pellets
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Ameisensäure, reagent grade, ≥95%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Natriumhydroxid, reagent grade, ≥98%, pellets (anhydrous)
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Dichlormethan, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%, contains 50-150 ppm amylene as stabilizer
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Natronlauge, 50% in H2O
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Acetonitril, ACS reagent, ≥99.5%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Borsäure, ACS reagent, ≥99.5%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Dichlormethan, anhydrous, ≥99.8%, contains 40-150 ppm amylene as stabilizer
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
2-Propanol, BioReagent, ≥99.5%, Molecular Biology
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Ameisensäure, ACS reagent, ≥96%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Acetonitril, anhydrous, 99.8%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Isopropylalkohol, meets USP testing specifications
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
2-Propanol, HPLC Plus, for HPLC, GC, and residue analysis, 99.9%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Natronlauge, BioUltra, Molecular Biology, 10 M in H2O
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Ammoniak -Lösung, 7 N in methanol
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
2-Propanol, anhydrous, 99.5%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Kaliumchlorid, ReagentPlus®, ≥99.0%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Natronlauge, 1.0 N, BioReagent, suitable for cell culture
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Acetonitril, suitable for HPLC, gradient grade, ≥99.9%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.