Merck
Human mesenchymal stromal cells suppress T-cell proliferation independent of heme oxygenase-1.
Cytotherapy (2015-01-18)
Seema R Patel, Ian B Copland, Marco A Garcia, Richard Metz, Jacques Galipeau
ZUSAMMENFASSUNG
Mesenchymal stromal cells deploy immune suppressive properties amenable for use as cell therapy for inflammatory disorders. It is now recognized that mesenchymal stromal cells necessitate priming with an inflammatory milieu, in particular interferon-γ, to exert augmented immunosuppressive effects. It has been recently suggested that the heme-catabolizing enzyme heme oxygenase-1 is an essential component of the mesenchymal stromal cell-driven immune suppressive response. Because mesenchymal stromal cells upregulate indoleamine 2,3-dioxygenase expression on interferon-γ priming and indoleamine 2,3-dioxygenase requires heme as a cofactor for optimal catabolic function, we investigated the potential antagonism of heme oxygenase-1 activity on indoleamine 2, 3-dioxygenase and the impact on mesenchymal stromal cell immune plasticity. We herein sought to evaluate the molecular genetic effect of cytokine priming on human mesenchymal stromal cell heme oxygenase-1 expression and its functional role in differentially primed mesenchymal stromal cells. Contrary to previous reports, messenger RNA and protein analyses demonstrated that mesenchymal stromal cells derived from normal subjects (n = 6) do not express heme oxygenase-1 at steady state or after interferon-γ, tumor necrosis factor-α, and/or transforming growth factor-β priming. Pharmacological inhibition of heme oxygenase-1 with the use of tin protoporphyrin did not significantly abrogate the ability of mesenchymal stromal cells to suppress T-cell proliferation in vitro. Overall, these results unequivocally demonstrate that under steady state and after cytokine priming, human mesenchymal stromal cells immunoregulate T-cell proliferation independent of heme oxygenase-1.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Dimethylsulfoxid, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Dimethylsulfoxid, ACS reagent, ≥99.9%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Dimethylsulfoxid, Molecular Biology
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Dimethylsulfoxid, suitable for HPLC, ≥99.7%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Dimethylsulfoxid, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Dimethylsulfoxid, ReagentPlus®, ≥99.5%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Dimethylsulfoxid, anhydrous, ≥99.9%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Dimethylsulfoxid, ≥99.5% (GC), suitable for plant cell culture
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Dimethylsulfoxid, BioUltra, Molecular Biology, ≥99.5% (GC)
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Fluoreszein-5(6)-isothiocyanat, BioReagent, suitable for fluorescence, mixture of 2 components, ≥90% (HPLC)
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Fluorescein-Isothiocyanat Isomer I, suitable for protein labeling, ≥90% (HPLC), powder
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Dimethylsulfid, ≥99%, FCC, FG
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Dimethylsulfoxid, meets EP testing specifications, meets USP testing specifications
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Dimethylsulfoxid, PCR Reagent
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Dimethylsulfid, ≥99%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
7-Aminoactinomycin D, ~97% (HPLC), powder
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
5(6)-Carboxyfluoreszeindiacetat-N-succinimidylester, BioReagent, suitable for fluorescence, ≥90% (HPLC)
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
meso-2,3-Dimercaptobernsteinsäure, ~98%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Fluoreszein-5(6)-isothiocyanat, ≥90% (HPLC)
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Dimethylsulfid, anhydrous, ≥99.0%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Fluorescein-Isothiocyanat Isomer I, ≥97.5% (HPLC)
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Supelco
Dimethylsulfoxid, analytical standard
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Dimethylsulfid, natural, ≥99%, FCC, FG
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Supelco
Dimethylsulfoxid, for inorganic trace analysis, ≥99.99995% (metals basis)
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
USP
Dimethylsulfoxid, United States Pharmacopeia (USP) Reference Standard
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Dimethylsulfoxid -Lösung, 50 wt. % in H2O
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Supelco
Dimethylsulfid, analytical standard
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
5-Carboxy-Fluoresceindiacetat N-Succinimidylester, suitable for fluorescence, ≥95.0% (HPLC)
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Dimethylsulfoxid, European Pharmacopoeia (EP) Reference Standard
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Dimethylsulfoxid, ≥99.6%, ReagentPlus®
Preise und Verfügbarkeit sind derzeit nicht verfügbar.