Merck
In vitro evaluation of transdermal patches of flurbiprofen with ethyl cellulose.
Acta poloniae pharmaceutica (2014-10-03)
Arfat Idrees, Nisar Ur Rahman, Zeeshan Javaid, Muhammad Kashif, Irfan Aslam, Khizar Abbas, Talib Hussain
ZUSAMMENFASSUNG
This study was aimed to determine effects of penetration enhancers and plasticizers on drug release from rationally designed formulations of flurbiprofen based transdermal drug delivery system. Matrix type transdermal patches were formulated with ethyl cellulose (EC) as a polymer by using plate casting method. The plasticizers such as propylene glycol (PG) and dibutyl phthalate (DBP), and enhancers such as Span 20, Tween 20, sodium lauryl sulfate (SLS), isopropyl myristate (IPM) and ethanol (EtOH) were formulated in different concentrations in the patches. Such different combinations of polymer with various enhancers and plasticizers in patches were evaluated for their effect on the physicochemical properties and drug release behavior of flurbiprofen. The drug release study was carried out by the paddle-over-disk method and permeation of drug was performed by Franz diffusion cell using rabbit skin. Patches having ethanol with ethyl cellulose showed more uniformity in the physical properties while the smoothness and clarity of patches containing sodium lauryl sulfate were not satisfactory. The drug release from patches followed Higuchi and Korsmeyer-Pappas model while maximum drug release was obtained by isopropyl myristate (903 microg). It was concluded that the patches having ethyl cellulose with isopropyl myristate and propylene glycol are more useful for transdermal patches of flurbiprofen.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Ethyl alcohol, Pure 200 proof, Molecular Biology
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, ACS reagent, ≥99.5%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Kaliumphosphat, ACS reagent, ≥99.0%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Natriumhydroxid, ACS reagent, ≥97.0%, pellets
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Chloroform, contains 100-200 ppm amylenes as stabilizer, ≥99.5%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Toluol, ACS reagent, ≥99.5%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Natriumhydroxid, reagent grade, ≥98%, pellets (anhydrous)
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Natriumdodecylsulfat, BioReagent, suitable for electrophoresis, Molecular Biology, ≥98.5% (GC)
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Chloroform, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%, contains amylenes as stabilizer
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Natronlauge, 50% in H2O
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, meets USP testing specifications
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Chloroform, contains ethanol as stabilizer, ACS reagent, ≥99.8%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Chloroform, suitable for HPLC, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Toluol, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Toluol, anhydrous, 99.8%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Natronlauge, BioUltra, Molecular Biology, 10 M in H2O
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Natriumdodecylsulfat, ≥99.0% (GC), dust-free pellets
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Ethyl alcohol, Pure 190 proof, for molecular biology
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Kaliumphosphat, powder, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.0%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Natronlauge, 1.0 N, BioReagent, suitable for cell culture
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Natriumdodecylsulfat, ACS reagent, ≥99.0%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Natriumhydroxid, BioXtra, ≥98% (acidimetric), pellets (anhydrous)
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Natriumdodecylsulfat -Lösung, BioUltra, Molecular Biology, 10% in H2O
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Natriumhydroxid, puriss., meets analytical specification of Ph. Eur., BP, NF, E524, 98-100.5%, pellets
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Propylenglycol, ≥99.5% (GC), FCC, FG
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Natriumdodecylsulfat, ReagentPlus®, ≥98.5% (GC)
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Natriumphosphat dibasisch Dodecahydrat, meets analytical specification of Ph. Eur., BP, E339, 98.5-102.5% (T)
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Phthalsäuredibutylester, 99%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.