Merck
Characterization of the xiamenmycin biosynthesis gene cluster in Streptomyces xiamenensis 318.
PloS one (2014-06-12)
Yong Yang, Ling Fu, Jinlong Zhang, Linghan Hu, Minjuan Xu, Jun Xu
ZUSAMMENFASSUNG
Xiamenmycin (1) is a prenylated benzopyran derivative with anti-fibrotic activity. To investigate the genetic basis of xiamenmycin biosynthesis, we performed genome mining in the xiamenmycin-producing Streptomyces xiamenensis wild-type strain 318 to identify a candidate gene cluster. The complete gene cluster, consisting of five genes, was confirmed by a series of gene inactivations and heterologous expression. Based on bioinformatics analyses of each gene and feeding experiments, we found that the structure of an intermediate xiamenmycin B (3) accumulated in a ximA inactivation mutant, allowing us to propose a biosynthetic pathway. All five of the genes in the pathway were genetically and biochemically characterized. XimA was biochemically characterized as an ATP-dependent amide synthetase, catalyzing an amide bond formation in the presence of ATP as the final step in Xiamenmycin biosynthesis. The Km value of XimA was determined to be 474.38 µM for the substrate xiamenmycin B. These studies provide opportunities to use genetic and chemo-enzymatic methods to create new benzopyran derivatives as potential therapeutic agents.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Chloramphenicol, ≥98% (HPLC)
Sigma-Aldrich
IPTG, ≥99% (TLC), ≤0.1% Dioxane
Sigma-Aldrich
Ampicillin, anhydrous, 96.0-102.0% (anhydrous basis)
Sigma-Aldrich
L-Threonin, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 99.0-101.0%
Sigma-Aldrich
4-Hydroxybenzoesäure, ReagentPlus®, ≥99%
Sigma-Aldrich
Chloramphenicol, BioReagent, suitable for plant cell culture
Sigma-Aldrich
4-Hydroxybenzoesäure, ReagentPlus®, 99%
Sigma-Aldrich
β-Nicotinamidadenin-Dinucleotidphosphat Hydrat
Sigma-Aldrich
L-Threonin, reagent grade, ≥98% (HPLC)
Supelco
4-Hydroxybenzoesäure, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Nalidixinsäure, ≥98%
SAFC
L-Threonin
Sigma-Aldrich
Isopropyl β-D-1-Thiogalactopyranosid, ≥99% (TLC)
Sigma-Aldrich
Isopropyl-β-D-thiogalactopyranosid -Lösung, ReadyMade IPTG solution for Blue-white screening
Supelco
Chloramphenicol, VETRANAL®, analytical standard
Supelco
Ampicillin
Sigma-Aldrich
Chloramphenicol, meets USP testing specifications
Sigma-Aldrich
L-Threonin, BioXtra, ≥99.5% (NT)
SAFC
Isopropyl β-D-1-Thiogalactopyranosid
Sigma-Aldrich
Ampicillin, meets USP testing specifications
Sigma-Aldrich
D-Threonin, ≥98% (TLC)
Sigma-Aldrich
Geranyl pyrophosphate ammonium salt, 1 mg/mL in methanol (:aqueous 10 mM NH4OH (7:3)), ≥95% (TLC)
Supelco
L-Threonin, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
β-Nicotinamidadenin-Dinucleotidphosphat Natriumsalz, pkg of 5 mg (per vial)
USP
Ampicillin, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
γ,γ-Dimethylallyl pyrophosphate triammonium salt, 1 mg/mL in methanol (:aqueous 10 mM NH4OH (7:3)), ≥90% (TLC)
Supelco
Nalidixinsäure, analytical standard
Chloramphenicol, European Pharmacopoeia (EP) Reference Standard
Ampicillin, anhydrous, European Pharmacopoeia (EP) Reference Standard
Supelco
L-Threonin, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland