Merck
  • A new and fast DLLME-CE method for the enantioselective analysis of zopiclone and its active metabolite after fungal biotransformation.
A new and fast DLLME-CE method for the enantioselective analysis of zopiclone and its active metabolite after fungal biotransformation.
Journal of pharmaceutical and biomedical analysis (2015-03-18)
Nayara Cristina Perez de Albuquerque, Cristiane Masetto de Gaitani, Anderson Rodrigo Moraes de Oliveira
ZUSAMMENFASSUNG
Zopiclone (ZO) is a chiral drug that undergoes extensive metabolism to N-desmethylzopiclone (N-Des-ZO) and zopiclone-N-oxide (N-Ox-ZO). Pharmacological studies have shown (S)-N-Des-ZO metabolite presents anxiolytic activity and a patent for this metabolite was requested for anxiety treatment and related disorders. In this context, biotransformation employing fungi may be a promising strategy to obtain N-Des-ZO. To perform the biotransformation study in this work, an enantioselective method based on capillary electrophoresis (CE) and dispersive liquid-liquid microextraction (DLLME) was developed. CE analyses were carried out in sodium phosphate buffer (pH 2.5; 50mmolL(-1)) containing 0.5% (w/v) carboxymethyl-β-CD, at a constant voltage of +25kV. DLLME was conducted using 2mL of liquid culture medium pH 9.5. Chloroform (100μL) and methanol (300μL) were employed as extraction and disperser solvent, respectively. After CE and DLLME optimization, the analytical method was fully validated. The method was linear over a concentration range of 90-6000ngmL(-1) for each ZO enantiomer (r>0.999) and 50-1000ngmL(-1) for each N-Des-ZO enantiomer (r>0.998). Absolute recovery of 51 and 82% was achieved for N-Des-ZO and ZO, respectively. The accuracy and precision results agreed with the EMA (European Medicines Agency) guideline, and so did the stability study. Application of the developed method in a biotransformation study was conducted in order to investigate the ability of fungi, belonging to the genus Cunninghamella, in metabolizing ZO chiral drug. Fungi Cunninghamella elegans ATCC 10028B and Cunninghamella echinulata var elegans ATCC 8688A demonstrated to be able to enantioselectively biotransform ZO to its active metabolite, N-Des-ZO. Therefore, the proposed goals of this work, i.e. a fast DLLME-CE method and an outstanding strategy to obtain N-Des-ZO, were successfully attained.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Acetonitril, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Ethyl alcohol, Pure 200 proof, Molecular Biology
Sigma-Aldrich
2-Propanol, suitable for HPLC, 99.9%
Sigma-Aldrich
Aceton, ACS reagent, ≥99.5%
Sigma-Aldrich
2-Propanol, ACS reagent, ≥99.5%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, ACS reagent, ≥99.5%
Sigma-Aldrich
Dichlormethan, suitable for HPLC, ≥99.8%, contains amylene as stabilizer
Sigma-Aldrich
N,N-Dimethylformamid, ACS reagent, ≥99.8%
Sigma-Aldrich
Aceton, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Acetonitril, HPLC Plus, ≥99.9%
Sigma-Aldrich
N,N-Dimethylformamid, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Dichlormethan, contains 40-150 ppm amylene as stabilizer, ACS reagent, ≥99.5%
Sigma-Aldrich
Natriumhydroxid, ACS reagent, ≥97.0%, pellets
Sigma-Aldrich
Trizma® Base, Primary Standard and Buffer, ≥99.9% (titration), crystalline
Sigma-Aldrich
Chloroform, contains 100-200 ppm amylenes as stabilizer, ≥99.5%
Sigma-Aldrich
Aceton, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%
Sigma-Aldrich
Natriumhydroxid, reagent grade, ≥98%, pellets (anhydrous)
Sigma-Aldrich
Phosphorsäure, ACS reagent, ≥85 wt. % in H2O
Sigma-Aldrich
Dichlormethan, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%, contains 50-150 ppm amylene as stabilizer
Sigma-Aldrich
N,N-Dimethylformamid, anhydrous, 99.8%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Chloroform, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%, contains amylenes as stabilizer
Sigma-Aldrich
Natronlauge, 50% in H2O
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, meets USP testing specifications
Sigma-Aldrich
Saccharose, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Acetonitril, ACS reagent, ≥99.5%
Sigma-Aldrich
Chloroform, contains ethanol as stabilizer, ACS reagent, ≥99.8%
Sigma-Aldrich
Chloroform, suitable for HPLC, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer