Merck
  • Design, synthesis, and local anti-inflammatory activity of 17β-carboxamide derivatives of glucocorticoids.
Design, synthesis, and local anti-inflammatory activity of 17β-carboxamide derivatives of glucocorticoids.
Archiv der Pharmazie (2014-08-28)
Vladimir Dobričić, Bojan Marković, Nikola Milenković, Vladimir Savić, Vesna Jaćević, Nemanja Rančić, Sote Vladimirov, Olivera Cudina
ZUSAMMENFASSUNG
Molecular docking studies were performed on 18 17β-carboxamide steroids in order to select compounds with potential local anti-inflammatory activity. These derivatives are amides of cortienic acids (obtained from hydrocortisone, prednisolone, and methylprednisolone) with methyl or ethyl esters of six amino acids. Interactions with the glucocorticoid receptor (GR), binding energies and ligand efficiency values of these compounds were compared with dexamethasone and cortienic acid obtained from prednisolone (inactive metabolite). On the basis of molecular docking studies, seven compounds were selected and their binding affinities for the GR were predicted by use of the exponential model created in this study. Subsequently, selected compounds were synthesized in good yields by use of modified N,N'-dicyclohexylcarbodiimide (DCC)/1-hydroxybenzotriazole (HOBt) coupling procedure. Finally, the local anti-inflammatory activity of the synthesized compounds was examined by use of the croton oil-induced ear edema test. In vivo evaluation of systemic side effects as well as in silico prediction of metabolism were performed on the derivative with the best local anti-inflammatory activity. The combination of molecular docking studies and the exponential model for the GR binding affinity prediction could be used as an in silico tool for the rational design of novel 17β-carboxamide steroids with potentially better biological profile than dexamethasone.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Essigsäure, glacial, ACS reagent, ≥99.7%
Sigma-Aldrich
N,N-Dimethylformamid, ACS reagent, ≥99.8%
Sigma-Aldrich
N,N-Dimethylformamid, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Essigsäure, glacial, ReagentPlus®, ≥99%
Sigma-Aldrich
Chloroform, contains 100-200 ppm amylenes as stabilizer, ≥99.5%
Sigma-Aldrich
N,N-Dimethylformamid, anhydrous, 99.8%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Chloroform, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%, contains amylenes as stabilizer
Sigma-Aldrich
Chloroform, contains ethanol as stabilizer, ACS reagent, ≥99.8%
Sigma-Aldrich
Chloroform, suitable for HPLC, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Essigsäure, glacial, ≥99.99% trace metals basis
Sigma-Aldrich
Essigsäure -Lösung, suitable for HPLC
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
N,N-Dimethylformamid, ReagentPlus®, ≥99%
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Chloroform, contains amylenes as stabilizer, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Chloroform, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
N,N-Dimethylformamid, Molecular Biology, ≥99%
Sigma-Aldrich
Chloroform, ACS spectrophotometric grade, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
L-Phenylalanin, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Sigma-Aldrich
Chloroform, ReagentPlus®, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
L-Alanin, ≥98% (TLC)
Sigma-Aldrich
Chloroform, anhydrous, ≥99%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, anhydrous, contains amylenes as stabilizer, ≥99%