Merck
  • Minimal amounts of dipalmitoylphosphatidylcholine improve aerosol performance of spray-dried temocillin powders for inhalation.
Minimal amounts of dipalmitoylphosphatidylcholine improve aerosol performance of spray-dried temocillin powders for inhalation.
International journal of pharmaceutics (2015-10-13)
Brieuc Cuvelier, Pierre Eloy, Cristina Loira-Pastoriza, Bernard Ucakar, Abdoul Aziz Sanogo, Christine Dupont-Gillain, Rita Vanbever
ZUSAMMENFASSUNG
Administration of antibiotics by inhalation can greatly improve drug targeting to the site of respiratory infections. In addition, dry powder inhalers are particularly convenient for the patients. The purposes of this study were to demonstrate the interest of pulmonary temocillin delivery to reach high temocillin concentrations locally in the lungs as well as to prepare a spray-dried temocillin powder for inhalation using a minimal amount of generally recognized as safe excipients. Intratracheal instillation of a temocillin solution allowed to reach higher and more sustained drug concentrations in the lungs than intravenous injection in mice, although a 10-fold lower temocillin dose was delivered intratracheally than systemically. A spray-dried powder of pure temocillin presented a fine particle fraction of 9% of the dose loaded in the inhaler. However, the incorporation of 0.5% to 20% of dipalmitoylphosphatidylcholine (DPPC) in the powder increased the fine particle fraction 4- to 5-fold. X-ray photoelectron spectroscopy and X-ray diffraction revealed that DPPC concentrated at the particle surface with its aliphatic chains laterally packed. The minimal amount of DPPC needed to improve the aerosol performance of temocillin supports the use of this excipient in the formulation of cohesive antibiotic powders for inhalation.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Chloroform, contains 100-200 ppm amylenes as stabilizer, ≥99.5%
Sigma-Aldrich
Acetonitril, ACS reagent, ≥99.5%
Sigma-Aldrich
Chloroform, contains ethanol as stabilizer, ACS reagent, ≥99.8%
Sigma-Aldrich
Acetonitril, anhydrous, 99.8%
Sigma-Aldrich
Chloroform, contains amylenes as stabilizer, ACS reagent, ≥99.8%
Sigma-Aldrich
Chloroform, ACS spectrophotometric grade, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, ReagentPlus®, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, anhydrous, ≥99%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
L-Alanin, ≥98% (TLC)
Sigma-Aldrich
Schwefel, powder, 99.98% trace metals basis
Sigma-Aldrich
Schwefel, 99.998% trace metals basis
Sigma-Aldrich
Chloroform, anhydrous, contains amylenes as stabilizer, ≥99%
Sigma-Aldrich
Chloroform, contains ethanol as stabilizer, meets analytical specification of BP, 99-99.4% (GC)
Sigma-Aldrich
L-Alanin, from non-animal source, meets EP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Sigma-Aldrich
Chloroform, biotech. grade, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Acetonitril, biotech. grade, ≥99.93%
Sigma-Aldrich
Acetonitril, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
L-Alanin, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
Acetonitril, suitable for DNA synthesis, ≥99.9% (GC)
Sigma-Aldrich
L-Alanin, ≥99%
Sigma-Aldrich
Sulforhodamin 101, Dye content ~95 %
Sigma-Aldrich
2,3-Dimercapto-1-propanol, ≥98% (iodometric)
Sigma-Aldrich
Acetonitril, ReagentPlus®, 99%
Sigma-Aldrich
Chloroform, ≥99%, PCR Reagent, contains amylenes as stabilizer
Sigma-Aldrich
Schwefel, flakes, ≥99.99% trace metals basis
Sigma-Aldrich
L-Alanin-12C3, 99.9 atom % 12C