Merck
  • Interferon regulatory factor 7 protects against vascular smooth muscle cell proliferation and neointima formation.
Interferon regulatory factor 7 protects against vascular smooth muscle cell proliferation and neointima formation.
Journal of the American Heart Association (2014-10-12)
Ling Huang, Shu-Min Zhang, Peng Zhang, Xiao-Jing Zhang, Li-Hua Zhu, Ke Chen, Lu Gao, Yan Zhang, Xiang-Jie Kong, Song Tian, Xiao-Dong Zhang, Hongliang Li
ZUSAMMENFASSUNG
Interferon regulatory factor 7 (IRF7), a member of the interferon regulatory factor family, plays important roles in innate immunity and immune cell differentiation. However, the role of IRF7 in neointima formation is currently unknown. Significant decreases in IRF7 expression were observed in vascular smooth muscle cells (VSMCs) following carotid artery injury in vivo and platelet-derived growth factor-BB (PDGF-BB) stimulation in vitro. Compared with non-transgenic (NTG) controls, SMC-specific IRF7 transgenic (IRF7-TG) mice displayed reduced neointima formation and VSMC proliferation in response to carotid injury, whereas a global knockout of IRF7 (IRF7-KO) resulted in the opposite effect. Notably, a novel IRF7-KO rat strain was successfully generated and used to further confirm the effects of IRF7 deletion on the acceleration of intimal hyperplasia based on a balloon injury-induced vascular lesion model. Mechanistically, IRF7's inhibition of carotid thickening and the expression of VSMC proliferation markers was dependent on the interaction of IRF7 with activating transcription factor 3 (ATF3) and its downstream target, proliferating cell nuclear antigen (PCNA). The evidence that IRF7/ATF3-double-TG (DTG) and IRF7/ATF3-double-KO (DKO) mice abolished the regulatory effects exhibited by the IRF7-TG and IRF7-KO mice, respectively, validated the underlying molecular events of IRF7-ATF3 interaction. These findings demonstrated that IRF7 modulated VSMC proliferation and neointima formation by interacting with ATF3, thereby inhibiting the ATF3-mediated induction of PCNA transcription. The results of this study indicate that IRF7 is a novel modulator of neointima formation and VSMC proliferation and may represent a promising target for vascular disease therapy.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Natriumchlorid, Molecular Biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Natriumchlorid -Lösung, 5 M in H2O, BioReagent, Molecular Biology
Sigma-Aldrich
Natriumchlorid, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Natriumchlorid, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Natriumchlorid -Lösung, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
L-Glutathion reduziert, suitable for cell culture, BioReagent, ≥98.0%, powder
Sigma-Aldrich
L-Glutathion reduziert, ≥98.0%
Sigma-Aldrich
Phenol -Lösung, Equilibrated with 10 mM Tris HCl, pH 8.0, 1 mM EDTA, BioReagent, Molecular Biology
Sigma-Aldrich
IPTG, ≥99% (TLC), ≤0.1% Dioxane
Sigma-Aldrich
Ethylendiamintetraessigsäure, 99.4-100.6%, powder
Sigma-Aldrich
Ethylendiamintetraessigsäure, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Natriumchlorid -Lösung, 5 M
Sigma-Aldrich
Phenol, puriss., ≥99.5% (GC), meets analytical specification of Ph. Eur., BP, USP, crystalline (detached)
Sigma-Aldrich
Ethylendiamintetraessigsäure -Lösung, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
SAFC
Natriumchlorid -Lösung, 5 M
Sigma-Aldrich
Chloroform, anhydrous, ≥99%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Ethylendiamintetraessigsäure, 99.995% trace metals basis
Sigma-Aldrich
Ethylendiamintetraessigsäure, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Chloroform, anhydrous, contains amylenes as stabilizer, ≥99%
Sigma-Aldrich
Phenol -Lösung, Saturated with 0.01 M citrate buffer, pH 4.3 ± 0.2, BioReagent, Molecular Biology
Sigma-Aldrich
Natriumchlorid, BioUltra, Molecular Biology, ≥99.5% (AT)
Sigma-Aldrich
Phenol, ≥99%
Sigma-Aldrich
Verflüssigtes Phenol, ≥89.0%
Sigma-Aldrich
Natriumchlorid, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Phenol, puriss. p.a., ACS reagent, reag. Ph. Eur., 99.0-100.5%
Sigma-Aldrich
Natriumchlorid -Lösung, BioUltra, Molecular Biology, ~5 M in H2O
Sigma-Aldrich
Natriumchlorid, 99.999% trace metals basis
Sigma-Aldrich
Phenol, Molecular Biology
Sigma-Aldrich
Isopropyl β-D-1-Thiogalactopyranosid, ≥99% (TLC)
Sigma-Aldrich
Isopropyl-β-D-thiogalactopyranosid -Lösung, ReadyMade IPTG solution for Blue-white screening