Merck
BMI1-RING1B is an autoinhibited RING E3 ubiquitin ligase.
Nature communications (2015-07-08)
Asad M Taherbhoy, Oscar W Huang, Andrea G Cochran
ZUSAMMENFASSUNG
Polycomb repressive complex 1 (PRC1) is required for ubiquitination of histone H2A lysine 119, an epigenetic mark associated with repression of genes important in developmental regulation. The E3 ligase activity of PRC1 resides in the RING1A/B subunit when paired with one of six PCGF partners. The best known of these is the oncogene BMI1/PCGF4. We find that canonical PRC1 E3 ligases such as PCGF4-RING1B have intrinsically very low enzymatic activity compared with non-canonical PRC1 RING dimers. The structure of a high-activity variant in complex with E2 (PCGF5-RING1B-UbcH5c) reveals only subtle differences from an earlier PCGF4 complex structure. However, two charged residues present in the modelled interface with E2-conjugated ubiquitin prove critical: in BMI1/PCGF4, these residues form a salt bridge that may limit efficient ubiquitin transfer. The intrinsically low activity of the PCGF4-RING1B heterodimer is offset by a relatively favourable interaction with nucleosome substrates, resulting in an efficient site-specific monoubiquitination.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration)
Sigma-Aldrich
Acetonitril, anhydrous, 99.8%
Sigma-Aldrich
Magnesiumchlorid -Lösung, Molecular Biology, 1.00 M±0.01 M
Sigma-Aldrich
Imidazol, ReagentPlus®, 99%
Sigma-Aldrich
DL-Dithiothreitol -Lösung, BioUltra, Molecular Biology, ~1 M in H2O
Sigma-Aldrich
Natriumchlorid, Molecular Biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Magnesiumchlorid, ≥98%
Sigma-Aldrich
MES, low moisture content, ≥99% (titration)
Sigma-Aldrich
Imidazol, ACS reagent, ≥99% (titration)
Sigma-Aldrich
Natriumchlorid -Lösung, 5 M in H2O, BioReagent, Molecular Biology
Sigma-Aldrich
Natriumchlorid, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Natriumchlorid, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Natriumchlorid -Lösung, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
L-Glutathion reduziert, suitable for cell culture, BioReagent, ≥98.0%, powder
Sigma-Aldrich
L-Glutathion reduziert, ≥98.0%
Supelco
DL-Dithiothreitol -Lösung, 1 M in H2O
Sigma-Aldrich
IPTG, ≥99% (TLC), ≤0.1% Dioxane
Sigma-Aldrich
Ethylendiamintetraessigsäure, 99.4-100.6%, powder
Sigma-Aldrich
BIS-TRIS, ≥98.0% (titration)
Sigma-Aldrich
Imidazol, Molecular Biology, ≥99% (titration)
Sigma-Aldrich
Ethylendiamintetraessigsäure, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Natriumchlorid -Lösung, 5 M
Sigma-Aldrich
Ethylendiamintetraessigsäure -Lösung, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
SAFC
Natriumchlorid -Lösung, 5 M
Sigma-Aldrich
Magnesiumchlorid, powder, <200 μm
Sigma-Aldrich
Imidazol, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
HEPES-Pufferlösung, 1 M in H2O
Sigma-Aldrich
Ethylendiamintetraessigsäure, 99.995% trace metals basis
Sigma-Aldrich
Ethylendiamintetraessigsäure, BioUltra, anhydrous, ≥99% (titration)