Merck
  • Hepatic transcriptional analysis in rats treated with Cassia occidentalis seed: involvement of oxidative stress and impairment in xenobiotic metabolism as a putative mechanism of toxicity.
Hepatic transcriptional analysis in rats treated with Cassia occidentalis seed: involvement of oxidative stress and impairment in xenobiotic metabolism as a putative mechanism of toxicity.
Toxicology letters (2014-06-29)
Gati Krushna Panigrahi, Ashish Yadav, Anuradha Yadav, Kausar M Ansari, Rajnish K Chaturvedi, Vipin M Vashistha, S Raisuddin, Mukul Das
ZUSAMMENFASSUNG
The present study was undertaken to investigate the effect of Cassia occidentalis (CO) seeds on the transcriptional expression patterns of mRNAs in rat liver by microarray analysis. The results indicated that exposure of CO (0.5%) seeds in diet to rats differentially regulated 60 transcripts belonging to various metabolic pathways including, oxidative stress, xenobiotic metabolism, carbohydrate metabolism, cell cycle, apoptosis etc. The expression of AKT1, CAT, SOD1, CYP1A1, CYP2B1, TGF-β, BAX, CREB1, JNK1 and IL-6 were validated by the qRT-PCR. In addition, involvement of oxidative stress was observed due to marked depletion of glutathione, increase in lipid peroxidation and modulation of antioxidant enzymes in hepatic tissue of rats treated with 0.5-2.0% CO in diet. Furthermore, significant decrease in the levels of Phase 1 (EROD, MROD and PROD) and Phase 2 (QR and GST) enzymes following 0.5-2.0% CO exposure indicates the impairment of xenobiotic metabolism and possible accumulation of toxic ingredients of the seeds in liver. Overall, the study predicts the involvement of multiple pathways and related biomolecules in CO induced hepatotoxicity and the data may be useful in formulating strategies for therapeutic interventions of suspected CO poisoning study cases.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Natriumdodecylsulfat, BioReagent, suitable for electrophoresis, Molecular Biology, ≥98.5% (GC)
Sigma-Aldrich
Glycin, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
Ammoniumpersulfat, ACS reagent, ≥98.0%
Sigma-Aldrich
Glycin, suitable for electrophoresis, ≥99%
Sigma-Aldrich
Natriumdodecylsulfat, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Diethylpyrocarbonat, 96% (NT)
Sigma-Aldrich
N,N,N′,N′-Tetramethylethylendiamin, BioReagent, suitable for electrophoresis, ≥99.0%
Sigma-Aldrich
Ammoniumpersulfat, Molecular Biology, suitable for electrophoresis, ≥98%
Sigma-Aldrich
Natriumdodecylsulfat, ACS reagent, ≥99.0%
Sigma-Aldrich
Natriumdodecylsulfat -Lösung, BioUltra, Molecular Biology, 10% in H2O
Sigma-Aldrich
L-Glutathion reduziert, suitable for cell culture, BioReagent, ≥98.0%, powder
Sigma-Aldrich
Ammoniumpersulfat, reagent grade, 98%
Sigma-Aldrich
L-Glutathion reduziert, ≥98.0%
Sigma-Aldrich
Acrylamid, suitable for electrophoresis, ≥99%
Sigma-Aldrich
Natriumdodecylsulfat, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Ethylendiamintetraessigsäure, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
N,N,N′,N′-Tetramethylethylendiamin, BioReagent, Molecular Biology, ≥99% (GC)
Sigma-Aldrich
Glycin, BioUltra, Molecular Biology, ≥99.0% (NT)
Sigma-Aldrich
Glycin, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, ≥98.5%
Sigma-Aldrich
Adenin, ≥99%
Sigma-Aldrich
Ethylendiamintetraessigsäure, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylendiamintetraessigsäure -Lösung, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Acrylamid, Molecular Biology, ≥99% (HPLC)
Sigma-Aldrich
Natriumdodecylsulfat, BioUltra, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
(Hydroxypropyl)methylcellulose, viscosity 2,600-5,600 cP, 2 % in H2O(20 °C)(lit.)
Sigma-Aldrich
Natriumdodecylsulfat -Lösung, BioUltra, 20% in H2O
Sigma-Aldrich
Ethylendiamintetraessigsäure Dinatriumsalz -Lösung, BioUltra, pH 8.0, ~0.5 M in H2O
Sigma-Aldrich
N,N,N′,N′-Tetramethylethylendiamin, ≥99.5%, purified by redistillation
Supelco
Natriumdodecylsulfat, dust-free pellets, suitable for electrophoresis, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Ethylendiamintetraessigsäure, 99.995% trace metals basis