Merck
Phenolic and carotenoid profiles and antiproliferative activity of foxtail millet.
Food chemistry (2014-12-23)
Li Zhen Zhang, Rui Hai Liu
ABSTRACT
Commonly consumed foxtail millet varieties Jingu28 and Jingu34 were compared in terms of phytochemical composition, antioxidant property, and antiproliferative activity. The cellular antioxidant activity (CAA) was evaluated based on HepG2 cell cultivation. Antiproliferative properties against HepG2 and MDA cell were assayed by methylene blue assay. Total phenolic content (TPC) was 78.79 and 114.22 mg gallic acid equiv/100 g DW in Jingu28 and Jingu34. Both varieties contained ferulic acid, chlorogenic acid, caffeic acid and p-coumaric acid, syringic acid. Xanthophylls and zeaxanthin were also detected. Peroxyl radical scavenging capacity of the foxtail millet were 228.13 (Jingu28) and 355.03 (Jingu34) μmol of vitamin C equiv/100 g, respectively. CAA values of the foxtail millet varieties ranged from 1.52 to 8.97 μmol quercetin equiv/100 g DW. The proliferation of MDA and HepG2 cancer cells were significantly inhibited in a dose-dependent manner after exposure to Jingu28 and Jingu34 extractions.
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Metanolo, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Ethyl alcohol, Pure 200 proof, Molecular Biology
Sigma-Aldrich
Acetone, ACS reagent, ≥99.5%
Sigma-Aldrich
Acido trifluoroacetico, ReagentPlus®, 99%
Sigma-Aldrich
Metanolo, ACS reagent, ≥99.8%
Sigma-Aldrich
Acido cloridrico, ACS reagent, 37%
Sigma-Aldrich
Acido trifluoroacetico, suitable for HPLC, ≥99.0%
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, ACS reagent, ≥99.5%
Sigma-Aldrich
Etilacetato, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetone, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Metanolo, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%
Sigma-Aldrich
Etilacetato, suitable for HPLC, ≥99.7%
Sigma-Aldrich
Sodio idrossido, ACS reagent, ≥97.0%, pellets
Sigma-Aldrich
Solfato di sodio, ACS reagent, ≥99.0%, anhydrous, granular
Sigma-Aldrich
Acetone, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%
Sigma-Aldrich
Sodio idrossido, reagent grade, ≥98%, pellets (anhydrous)
Sigma-Aldrich
Solfato di sodio, ACS reagent, ≥99.0%, anhydrous, powder
Sigma-Aldrich
Metanolo, HPLC Plus, ≥99.9%
Sigma-Aldrich
Acetone, Laboratory Reagent, ≥99.5%
Sigma-Aldrich
Idrossido di sodio, 50% in H2O
Sigma-Aldrich
Esano, suitable for HPLC, ≥97.0% (GC)
Sigma-Aldrich
Acido cloridrico, ACS reagent, 37%
Sigma-Aldrich
ter-butil metil etere, suitable for HPLC, ≥99.8%
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, meets USP testing specifications
Sigma-Aldrich
Etilacetato, HPLC Plus, for HPLC, GC, and residue analysis, 99.9%
Sigma-Aldrich
Acetonitrile, ACS reagent, ≥99.5%
Sigma-Aldrich
Cloruro di idrogeno, 4.0 M in dioxane
Sigma-Aldrich
Esano, ReagentPlus®, ≥99%