Merck
  • Association efficiency of three ionic forms of oxytetracycline to cationic and anionic oil-in-water nanoemulsions analyzed by diafiltration.
Association efficiency of three ionic forms of oxytetracycline to cationic and anionic oil-in-water nanoemulsions analyzed by diafiltration.
Journal of pharmaceutical sciences (2015-01-06)
Sandra L Orellana, Cesar Torres-Gallegos, Rodrigo Araya-Hermosilla, Felipe Oyarzun-Ampuero, Ignacio Moreno-Villoslada
ABSTRACT
The association efficiency of oxytetracycline (OTC) to pharmaceutical available, ionic oil-in-water nanoemulsions is studied. Theoretical mathematical developments allowed us to differentiate by diafiltration (DF) between thermodynamically and kinetically controlled binding of the drug to the nanoemulsions, and relate these important magnitudes to the association efficiency. The nanoemulsions have been prepared by the solvent displacement technique in the presence of cationic and anionic surfactants. The resulting nanoemulsions were stable at 4°C and 25°C for 60 days, have a size of ∼ 200 nm, showing polydispersity indexes ranging between 0.11 and 0.23, and present zeta potentials ranging between -90 and +60 mV, depending on the charge of the surfactants used. The zeta potential of the nanoemulsions influenced the interaction with OTC, having three ionic forms at different pH, namely, cationic, zwitterionic, and anionic. DF proved to be a powerful tool for the quantification of the drug association efficiency, achieving values up to 84%. Furthermore, this technique allowed obtaining different values of the drug fractions reversibly bound (11%-57%) and irreversibly bound (10%-40%) to the nanoemulsions depending on the surfactants used and pH. These findings may be useful for the development of new drug delivery systems, and as routine assays in academia and pharmaceutical industries.
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto
Sigma-Aldrich
Ethyl alcohol, Pure 200 proof, Molecular Biology
Sigma-Aldrich
Acetone, ACS reagent, ≥99.5%
Sigma-Aldrich
Acido cloridrico, ACS reagent, 37%
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetone, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Sodio idrossido, ACS reagent, ≥97.0%, pellets
Sigma-Aldrich
Acetone, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%
Sigma-Aldrich
Sodio idrossido, reagent grade, ≥98%, pellets (anhydrous)
Sigma-Aldrich
Sodio dodecil solfato, BioReagent, suitable for electrophoresis, Molecular Biology, ≥98.5% (GC)
Sigma-Aldrich
Acetone, Laboratory Reagent, ≥99.5%
Sigma-Aldrich
Idrossido di sodio, 50% in H2O
Sigma-Aldrich
Acido cloridrico, ACS reagent, 37%
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, meets USP testing specifications
Sigma-Aldrich
Cloruro di idrogeno, 4.0 M in dioxane
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
Sigma-Aldrich
Idrossido di sodio, BioUltra, Molecular Biology, 10 M in H2O
Sigma-Aldrich
Esadeciltrimetilammonio bromuro, ≥98%
Sigma-Aldrich
Sodio dodecil solfato, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Idrossido di sodio, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Acido cloridrico, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Sodio dodecil solfato, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodio idrossido, BioXtra, ≥98% (acidimetric), pellets (anhydrous)
Sigma-Aldrich
Sodio dodecil solfato, BioUltra, Molecular Biology, 10% in H2O
Sigma-Aldrich
Sodio idrossido, puriss., meets analytical specification of Ph. Eur., BP, NF, E524, 98-100.5%, pellets
Sigma-Aldrich
Esadeciltrimetilammonio bromuro, Molecular Biology, ≥99%
Sigma-Aldrich
Acido cloridrico, meets analytical specification of Ph. Eur., BP, NF, fuming, 36.5-38%
Sigma-Aldrich
Acido cloridrico, 37 wt. % in H2O, 99.999% trace metals basis
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
Acetone, suitable for HPLC, ≥99.8%
Sigma-Aldrich
Acetone, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.5% (GC)